Skip to Content
Merck

Skip To

ISOTS4R-EP

Eppendorf® IsoTherm-System®

IsoRack, set of 4 racks, for 1.5 mL/2.0 mL vessels

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
41102404
Manufacturer/tradename:
Eppendorf® 022510169(NA)
Feature:
for 1.5 mL/2.0 mL vessels
Packaging:
set of 4 racks
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


feature

for 1.5 mL/2.0 mL vessels

packaging

set of 4 racks

manufacturer/tradename

Eppendorf® 022510169(NA)

General description

The IsoTherm System specializes in cooling samples effectively and constantly at -21 °C or 0 °C over many hours. It consists of the following components: the IsoRack, IsoPack and IsoSafe. It can be used as an enzyme bar to prevent enzymes from becoming inactive as well as for PCR sample preparation, sample transport and slow thawing of frozen samples.
  • Transport and slow thawing of deep-frozen samples
  • Holds 24 tubes (Safe-Lock 0.5 mL/1.5 mL/2.0 mL)
  • IsoPack (cold pack), white for 0 °C and blue for -21 °C, cooled to refrigeration temperature in freezer compartment
  • Isothermal times of < 6 h (0 °C, white) and < 3 h (-21 °C, blue)
  • No more melting ice baths (danger of contamination)
  • All system components can be used separately
  • Racks stackable, autoclavable, markable, floatable and can be centrifuged in microtiter-plate rotor

Legal Information

Eppendorf is a registered trademark of Eppendorf SE
IsoTherm-System is a registered trademark of Eppendorf SE


Still not finding the right product?

Explore all of our products under Eppendorf® IsoTherm-System®




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
EP022510169-4EA04061835297665