Skip to Content
Merck
All Photos(1)

Documents

HPA040735

Sigma-Aldrich

Anti-RASSF1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-123F2, Anti-NORE2A, Anti-RDA32, Anti-REH3P21, Anti-Ras association (RalGDS/AF-6) domain family member 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

KFTCHYRCRALVCLDCCGPRDLGWEPAVERDTNVDEPVEWETPDLSQAEIEQKIKEYNAQINSNLFMSLNKDG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RASSF1(11186)

General description

The Ras association domain family 1 A (RASSF1A) is a tumor suppressor gene that spans around 1859bp and has 6 exons (1α, 2αβ, 3, 4, 5, 6). The C terminal region of RASSF1A has a Salvador-RASSF1A-Hippo (SARAH) domain and ATM (ataxia telangiectasia mutant) kinase phosphorylation sites. The terminal region has high cysteine resides. This gene is located on human chromosome 3p21.3.

Immunogen

Ras association (RalGDS/AF-6) domain family member 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The Ras association domain family 1 A (RASSF1A) regulates the cell cycle, mitosis and maintains the stability of microtubules. The terminal region that is rich in cysteine residues plays a major role in RASSF1A-mediated cell apoptosis. It participates in several molecular pathways like Ras/PI3K/AKT, Ras/RAF/MEK/ERK, Hippo pathways and β-catenin signaling pathway. The RASSF1A protein can prevent accumulation of cyclin D1 and stimulate cell cycle arrest.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST81790

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

RASSF1A promoter methylation in high-grade serous ovarian cancer: a direct comparison study in primary tumors, adjacent morphologically tumor cell-free tissues and paired circulating tumor DNA.
Giannopoulou L, et al.
Oncotarget, 8(13), 21429?21443-21429?21443 (2017)
Ras-association domain family 1 isoform A (RASSF1A) gene polymorphism rs1989839 is associated with risk and metastatic potential of osteosarcoma in young Chinese individuals: a multi-center, case-control study.
Xu H, et al.
Medical Science Monitor : International Medical Journal of Experimental and Clinical Research, 22, 4529-4529 (2016)
The relationship between RASSF1A gene promoter methylation and the susceptibility and prognosis of melanoma: A meta-analysis and bioinformatics.
Shao C, et al.
PLoS ONE, 12(2), e0171676-e0171676 (2017)
Maria Chatzifrangkeskou et al.
The EMBO journal, 38(16), e101168-e101168 (2019-08-16)
Nuclear actin participates in many essential cellular processes including gene transcription, chromatin remodelling and mRNA processing. Actin shuttles into and out the nucleus through the action of dedicated transport receptors importin-9 and exportin-6, but how this transport is regulated remains

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service