Skip to Content
Merck
All Photos(6)

Key Documents

HPA011038

Sigma-Aldrich

Anti-NECTIN3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CD113, Anti-CDw113, Anti-DKFZP566B0846, Anti-PPR3, Anti-PVRL3, Anti-PVRR3, Anti-nectin-3, Anti-CD113 antigen, Anti-Nectin-3, Anti-Poliovirus receptor-related protein 3 precursor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

IHGKSSQTVAVHHPQYGFSVQGEYQGRVLFKNYSLNDATITLHNIGFSDSGKYICKAVTFPLGNAQSSTTVTVLVEPTVSLIKGPDSLIDGGNETVAAICIAATGKPVAHIDWEGDLGEMESTTTSF

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PVRL3(25945)

General description

PVRL3 (poliovirus receptor-related 3) belongs to nectin family, which is a family of immunoglobulin-like molecules, and are involved in cell adhesion. This family contains four members namely, PVRL1, PVRL2, PVRL3 and PVRL4. These are ubiquitously expressed and have multiple isoforms due to alternative splicing. PVRL3 is also called nectin-3 and has three isoforms called nectin-3α, -3β and -3δ. These variants differ in their transmembrane and cytosolic domains, but have identical extracellular regions. This is the only nectin to be expressed by T-cells.

Immunogen

nectin cell adhesion molecule 3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PVRL3 (poliovirus receptor-related 3) is a cell-cell adhesion molecule. It forms trans-dimers with nectin-1, and also interacts with afadin to form cell-cell adhesions in non-epithelial cells, which lack tight junctions. PVRL3 present on lymphocytes interacts with nectin-2 expressed on vascular endothelial cells. This interaction is essential for leukocyte migration and extravasation. This is the first member of nectin family which plays a role in determining the ocular phenotype in humans, and is involved in the development of mammalian lens and ciliary body. Membranous expression of PVRL3 without E-cadherin co-localization in cancers, usually predicts poor prognosis and overall survival of patients. The expression of this protein is deregulated in breast cancer, and it might act as tumor suppressor and prevent metastasis in breast cancer cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72277

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tracey A Martin et al.
PloS one, 8(12), e82696-e82696 (2014-01-05)
Nectins are a family of integral protein molecules involved in the formation of functioning Adherens and Tight Junctions (TJ). Aberrant expression is associated with cancer progression but little is known how this effects changes in cell behaviour. This study aimed
Elisabeth Devilard et al.
PloS one, 8(10), e77424-e77424 (2013-10-12)
Lymphocyte trafficking and migration through vascular endothelial cells (ECs) in secondary lymphoid tissues is critical for immune protection. In the present study, we investigate the role of nectin cell adhesion molecules for the migration of lymphocytes through ECs. Nectins are
Salil A Lachke et al.
Human genetics, 131(2), 235-250 (2011-07-20)
We describe a male patient (patient DGAP113) with a balanced translocation, 46,XY,t(1;3)(q31.3;q13.13), severe bilateral congenital cataracts, CNS abnormalities and mild developmental delay. Fluorescence in situ hybridization (FISH) and suppression PCR demonstrated that the chromosome 3 breakpoint lies ~515 kb upstream of
Henry Haeberle et al.
Neoplasia (New York, N.Y.), 14(7), 666-669 (2012-08-21)
High-resolution image guidance for resection of residual tumor cells would enable more precise and complete excision for more effective treatment of cancers, such as medulloblastoma, the most common pediatric brain cancer. Numerous studies have shown that brain tumor patient outcomes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service