Skip to Content
Merck
All Photos(5)

Key Documents

HPA010703

Sigma-Aldrich

Anti-DNASE1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DNL1, Anti-deoxyribonuclease I

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€505.00

About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

YVRPSQWSSIRLWTSPTFQWLIPDSADTTATPTHCAYDRIVVAGMLLRGAVVPDSALPFNFQAAYGLSDQLAQAISDHY

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DNASE1(1773)

General description

The enzyme is Ca2+ and Mg2+ dependent. It is synthesized in the pancreas and stored in zymogen granules. It has been used to reduce the viscosity of cystic fibrosis sputum.

Immunogen

deoxyribonuclease I recombinant protein epitope signature tag (PrEST)

Application

Anti-DNASE1 antibody produced in rabbit has been used for immunostaining of DNAseI.
Anti-DNASE1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

DNase I is an endonuclease that hydrolyzes double-stranded or single-stranded DNA preferentially at sites adjacent to pyrimidine nucleotides to produce a complex mixture of 5 ′-phosphate mononucleotides and oligonucleotides. Mutation in this gene is associated with systemic lupus erythematosus (SLE), an autoimmune disease.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71906

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Panneer et al.
Lupus, 22(10), 996-1000 (2013-08-22)
Systemic lupus erythematosus (SLE) is a multisystem disorder in which defective apoptotic clearance is considered to be an important factor in pathogenesis. DNAse I is associated with disposal of apoptotic nuclear debris. The defective enzyme production due to +2373 A to
S Shak et al.
Proceedings of the National Academy of Sciences of the United States of America, 87(23), 9188-9192 (1990-12-01)
Respiratory distress and progressive lung destruction in cystic fibrosis can be attributed to bacterial persistence and the accumulation of viscous purulent secretions in the airways. More than 30 yr ago it was suggested that the large amounts of DNA in
Goetz Parsiegla et al.
Biochemistry, 51(51), 10250-10258 (2012-12-12)
Recombinant human DNase I (Pulmozyme, dornase alfa) is used for the treatment of cystic fibrosis where it improves lung function and reduces the number of exacerbations. The physiological mechanism of action is thought to involve the reduction of the viscoelasticity
Snehal Sonawane et al.
Investigative ophthalmology & visual science, 53(13), 8253-8263 (2012-11-22)
We determined whether nucleases are deficient in the tear fluid of dry eye disease (DED) patients, and whether this causes extracellular DNA (eDNA) and neutrophil extracellular trap (NET) accumulation in the precorneal tear film, thus causing ocular surface inflammation. Exfoliated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service