Skip to Content
Merck
All Photos(10)

Documents

HPA000792

Sigma-Aldrich

Anti-CKAP4 antibody produced in rabbit

enhanced validation

Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-63 kDa membrane protein antibody produced in rabbit, Anti-Cytoskeleton-associated protein 4 antibody produced in rabbit, Anti-p63 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, mouse, human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

LKDLSDGIHVVKDARERDFTSLENTVEERLTELTKSINDNIAIFTEVQKRSQKEINDMKAKVASLEESEGNKQDLKALKEAVKEIQTSAKSREWDMEALRSTLQTMES

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CKAP4(10970)

Looking for similar products? Visit Product Comparison Guide

Immunogen

Cytoskeleton-associated protein 4 recombinant protein epitope signature tag (PrEST)

Application

Anti-CKAP4 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Cytoskeleton-associated protein 4 is a protein encoded by the CKAP4 gene in humans. It is a 63kDa membrane protein and is a resident protein of a membrane network interposed in between rough ER and golgi apparatus. The protein is a transcription factor that plays pivotal roles in epidermal barrier formation and in embryonic development. CKAP4 is found to be overexpressed in intrahepatic cholangiocellular carcinoma (ICC) and is associated with tumor size, distant metastasis and lymph node metastasis. It may act as a prognostic marker for ICC patients.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76131

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manbo Cai et al.
Experimental biology and medicine (Maywood, N.J.), 248(12), 1085-1094 (2023-05-20)
Cytoskeleton-associated protein 4 (CKAP4) acts as a key transmembrane protein that connects the endoplasmic reticulum (ER) to microtubule dynamics. Researchers have not examined the roles of CKAP4 in nasopharyngeal carcinoma (NPC). The study aimed at evaluating the prognostic value and
Min-hong Li et al.
Cancer letters, 337(2), 248-253 (2013-05-15)
The objective of the study was to investigate the clinical significance of CKAP4 in intrahepatic cholangiocellular carcinoma (ICC). CKAP4 expression was determined in a cohort containing 173 cases of ICC patients. We found that CKAP4 was overexpressed in the majority
Stefano Rufini et al.
Biochemical pharmacology, 82(10), 1262-1268 (2011-08-09)
The transcription factor p63 plays pivotal roles in epidermal barrier formation and in embryonic development. The protein structures of TAp63 and ΔNp63α isoforms include a C-terminal steril alpha-motif (SAM) involved in protein-protein interaction. Identification of p63 SAM domain interactors could
A Schweizer et al.
Journal of cell science, 104 ( Pt 3), 685-694 (1993-03-01)
The recently identified 63 kDa membrane protein, p63, is a resident protein of a membrane network interposed in between rough ER and Golgi apparatus. To characterize p63 at the molecular level a 2.91 kb cDNA encoding p63 has been isolated
Shuang-Xi Li et al.
Cancer, 120(10), 1520-1531 (2014-05-28)
The functions of cytoskeleton-associated membrane protein 4 (CKAP4), one kind of type II transmembrane protein, are associated with the palmitoyl acyltransferase DHHC2. The objective of the current study was to investigate CKAP4/DHHC2 expression and its prognostic significance in patients with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service