Skip to Content
Merck
All Photos(1)

Key Documents

H1142

Sigma-Aldrich

15(S)-Hydroxy-(5Z,8Z,11Z,13E)-eicosatetraenoic acid

~150 μg/mL in ethanol, ≥97.0% (HPLC)

Synonym(s):

(15S,5Z,8Z,11Z,13E)-15-Hydroxyeicosatetraenoic acid, (S)-15-HETE, 15(S)-HETE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32O3
CAS Number:
Molecular Weight:
320.47
Beilstein:
2470466
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97.0% (HPLC)

concentration

~150 μg/mL in ethanol

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCC[C@H](O)\C=C\C=C/C\C=C/C\C=C/CCCC(O)=O

InChI

1S/C20H32O3/c1-2-3-13-16-19(21)17-14-11-9-7-5-4-6-8-10-12-15-18-20(22)23/h4-5,8-11,14,17,19,21H,2-3,6-7,12-13,15-16,18H2,1H3,(H,22,23)/b5-4-,10-8-,11-9-,17-14+/t19-/m0/s1

InChI key

JSFATNQSLKRBCI-VAEKSGALSA-N

Biochem/physiol Actions

Metabolite of arachidonic acid via the 15-lipoxygenase pathway that may be involved in pulmonary anti-inflammatory responses through the inhibition of 5-lipoxygenase. 15(S)-HETE suppresses the incorporation of thymidine and biosynthesis of prostaglandin E2 in tumor cell cultures. Treatment of colon cancer cells with 15(S)-HETE inhibited cell proliferation and induced apoptosis, which was preceded by an increase in TGF-beta-inducible early gene (TIEG) and a decrease in Bcl-2.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Profita et al.
American journal of respiratory cell and molecular biology, 20(1), 61-68 (1998-12-31)
15(S)-Hydroxyeicosatetraenoic acid (15[S]-HETE) is a 15-lipoxygenase (15-LO) metabolite that may play an important role in different pulmonary diseases. 15-HETE is synthesized by different epithelial cells and may be subsequently incorporated into cellular phospholipids. We studied the role of interleukin-4 (IL-4)
M Profita et al.
American journal of physiology. Cell physiology, 279(4), C1249-C1258 (2000-09-26)
We evaluated the levels of 15(S)-hydroxyeicosatetraenoic acid [15(S)-HETE] and the expression of 15-lipoxygenase (15-LO) mRNA in induced sputum obtained from 10 control and 15 chronic bronchitis subjects. 15(S)-HETE was evaluated by reverse phase high-performance liquid chromatography separation followed by specific
Eicosanoids in inflammatory skin diseases.
K Fogh et al.
Prostaglandins & other lipid mediators, 63(1-2), 43-54 (2000-12-05)
Cui Ma et al.
Hypertension (Dallas, Tex. : 1979), 58(4), 679-688 (2011-08-24)
We have found that 15-hydroxyeicosatetraenoic acid (15-HETE) induced by hypoxia was an important mediator in the regulation of hypoxic pulmonary hypertension, including the pulmonary vasoconstriction and remodeling. However, the underlying mechanisms of the remodeling induced by 15-HETE are poorly understood.
Li-Xing Tian et al.
Cellular immunology, 349, 104047-104047 (2020-02-06)
The polarization of macrophages is critical to inflammation and tissue repair, with unbalanced macrophage polarization associated with critical dysfunctions of the immune system. Cytochrome P450 1A1 (CYP1A1) is a hydroxylase mainly controlled by the inflammation-limiting aryl hydrocarbon receptor (AhR), which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service