GE29-0181-84
HiPrep™ Phenyl High Performance16/10
Cytiva 29-0181-84, pack of 1 column
About This Item
Recommended Products
ligand
phenyl
packaging
pack of 1 column
manufacturer/tradename
Cytiva 29-0181-84
parameter
1-5 ml/min flow rate
44 psi
bed size
16 mm × 100 mm
column I.D.
16 mm
matrix
6% cross-linked agarose
particle size
24-44 μm
avg. part. size
34 μm
cleaning
2-14
working range
3-13
suitability
suitable for bioprocess medium (BioProcess medium.)
suitable for bioprocess medium
Related Categories
General description
Features and Benefits
- Aromatic HIC resin for intermediate purification and polishing.
- Selectivity similar to Phenyl Sepharose 6 Fast Flow (Low Sub) but higher resolution due to the smaller 34 µm bead size.
- Provides fast, easy high-capacity separations in a convenient format.
Storage and Stability
Analysis Note
Other Notes
Legal Information
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
3 - Flammable liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service