Skip to Content
Merck
All Photos(1)

Key Documents

GE28-9365-48

HiPrep Octyl Fast Flow 16/10

Cytiva 28-9365-48

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
13111000
NACRES:
NA.56

ligand

octyl

packaging

pkg of 1 ea

manufacturer/tradename

Cytiva 28-9365-48

parameter

1.5 bar (22 psi) (Over the Packed Bed During Operation)
2 ml/min - 10 ml/min flow rate
22 psi

bed volume

20 mL

column I.D.

16 mm

matrix

4% cross-linked agarose

particle size

45-165 μm

average diameter

90 μm

cleaning

2-14

working range

3-13

suitability

suitable for bioprocess medium

General description

HiPrep Octyl FF (High Sub) 16/10, are prepacked Octyl Sepharose 4 Fast FLow columns ready to use for hydrophobic interaction chromatography (HIC). The columns are designed for initial and intermediate purification, removing bulk impurities.

Features and Benefits

  • 20 mL HIC columns provide fast, preparative separation of Proteins and other biomolecules.
  • Provide high capacity, excellent flow properties, and reliable, reproducible separations.
  • Octyl Sepharose 4 Fast FLow is a well-established, standard aliphatic HIC medium for capture and intermediate purification.
  • Compatible with simple pump-based configurations or chromatography system such as ÄKTA® design.
  • The octyl derivative adds a different and complementary selectivity to the Sepharose Fast FLow HIC product family.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Other Notes

The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used.

Legal Information

HiPrep is a trademark of Cytiva
Sepharose is a trademark of Cytiva
ÄKTA is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

This page covers detailed information on cleaning procedures and recommended flow for column cleaning.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service