Skip to Content
Merck
All Photos(1)

Key Documents

F0627

Sigma-Aldrich

D-Fructose 1,6-bisphosphate barium salt

practical grade, 70-80%, amorphous powder

Synonym(s):

D(+)Fructofuranose 1,6-diphosphate, Harden-Young ester, Hexose diphosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H12O12P2Ba
CAS Number:
Molecular Weight:
475.43
UNSPSC Code:
12352200

grade

practical grade

Assay

70-80%

form

amorphous powder

Preparation Note

Prepared by a modification of the procedure of Neuberg, C., J. Am. Chem. Soc., 64, 2722 (1942).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service