Skip to Content
Merck
All Photos(1)

Documents

EHU099351

Sigma-Aldrich

MISSION® esiRNA

targeting human CDK1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCATGGGGATTCAGAAATTGATCAACTCTTCAGGATTTTCAGAGCTTTGGGCACTCCCAATAATGAAGTGTGGCCAGAAGTGGAATCTTTACAGGACTATAAGAATACATTTCCCAAATGGAAACCAGGAAGCCTAGCATCCCATGTCAAAAACTTGGATGAAAATGGCTTGGATTTGCTCTCGAAAATGTTAATCTATGATCCAGCCAAACGAATTTCTGGCAAAATGG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiajia Li et al.
Biochemical and biophysical research communications, 523(2), 434-440 (2019-12-27)
Ovarian cancer is the most lethal gynecological malignancy, but the mechanisms of ovarian cancer progression and cisplatin resistance remain unclear. Emerging evidence suggested that ubiquitin-conjugating enzyme E2C (UBE2C) was highly expressed in a variety of tumors and acted as an
Masanori Saito et al.
Scientific reports, 6, 20622-20622 (2016-02-11)
Skeletal development is tightly regulated through the processes of chondrocyte proliferation and differentiation. Although the involvement of transcription and growth factors on the regulation of skeletal development has been extensively studied, the role of cell cycle regulatory proteins in this
Gabriel Kollarovic et al.
Aging, 8(1), 158-177 (2016-02-03)
Excessive DNA damage can induce an irreversible cell cycle arrest, called senescence, which is generally perceived as an important tumour-suppressor mechanism. However, it is unclear how cells decide whether to senesce or not after DNA damage. By combining experimental data
Tatyana S Nekova et al.
Cell cycle (Georgetown, Tex.), 15(23), 3203-3209 (2016-11-11)
Small molecule inhibitors targeting CDK1/CDK2 have been clinically proven effective against a variety of tumors, albeit at the cost of profound off target toxicities. To separate potential therapeutic from toxic effects, we selectively knocked down CDK1 or CDK2 in p53
Chen Sai et al.
International heart journal, 60(2), 374-383 (2019-02-13)
Atrial fibrillation has caused severe burden for people worldwide. Differentiation of fibroblasts into myofibroblasts, and consequent progress in atrial structural remodeling have been considered the basis for persistent atrial fibrillation, yet little is known about the molecular mechanisms underlying the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service