DWP962000W8-EP
Eppendorf® Deepwell Plate 96/2000μL
PCR Clean, volume 2000 μL, white border with clear wells, pkg of 80 plates (10 bags x 8 plates)
About This Item
polypropylene
round bottom wells
white border with clear wells
Recommended Products
material
colorless wells
polypropylene
round bottom wells
feature
PCR Clean
white border with clear wells
packaging
pkg of 80 plates (10 bags x 8 plates)
manufacturer/tradename
Eppendorf® 951033600
availability
available only in North America
technique(s)
PCR: suitable
size
96 well
volume
2000 μL
working volume
2 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Features and Benefits
- OptiTrack® matrix: up to 30 % faster sample identification and fewer pipetting errors via high-contrast alphanumeric labeling
- RecoverMax® well design: Optimized well geometry for maximum sample recovery and excellent mixing properties
- Minimal remaining volume and high uniformity from well to well
- Raised well rims and smooth surface for reliable closing, even with heat sealing
- Easy and reliable stacking
- g-Safe®: exceptional centrifugation stability up to 6,000 × g for faster protocols and improved sample quality
- High-quality polypropylene guarantees high resistance to chemicals, mechanical stress and temperature extremes
- Manufactured without slip agents, plasticizers, and biocides – Materials which could have a negative effect on bioassays
- Autoclavable (121?°C, 20?min)
- Available with barcode. Additional details available at: www.eppendorf.com/barcode
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service