CLS6795171
Corning® Scholar® PC-171 magnetic Stirrer
top plate size 5 in. × 5 in., AC/DC input 120 V AC
Synonym(s):
stir plate, stirrer
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41103800
Recommended Products
material
platform (abs plastic)
measuring range
100-1600 rpm
top plate size
5 in. × 5 in.
weight
18 kg
Related Categories
General description
The 5 x 5 inch plastic top Scholar PC-171 Stirrer is compact and provides vigorous stirring with an alternating current motor for longer life and rugged construction. It operates on 120V/60Hz and is UL and CUL approved to meet needs in North America and other locations with similar requirements. -Space saving design and light weight. -UL, CUL and CE approved. -1 x 3/8 inch stir bar included with each stirrer
Legal Information
Corning is a registered trademark of Corning, Inc.
Scholar is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service