SK-N-DZ
94092305, human nerve (bone marrow metastasis), Neuronal
About This Item
Recommended Products
product name
SK-N-DZ, 94092305
biological source
human nerve (bone marrow metastasis)
growth mode
Adherent
karyotype
44XX, monosomic for 10, 11, 13, 14, 19. Both copies of chromosome 2 missing in each cell.
morphology
Neuronal
products
p53, moderate MDR-1 expression
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
metastasis
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Cell Line Description
Application
DNA Profile
CSF1PO: 12
D13S317: 8,11
D16S539: 9,11
D5S818: 12
D7S820: 12,13
THO1: 6,9
TPOX: 8
vWA: 16,18
Culture Medium
Subculture Routine
Other Notes
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
DNA, RNA, cDNA derived from ECACC mammalian cell lines allow screening for genes or expression patterns to identify lines most suitable for specific research.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service