Skip to Content
Merck
All Photos(1)

Key Documents

COR-L51

92031916, human lung, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

COR-L51, 92031916

biological source

human lung

growth mode

Suspension

karyotype

Not specified

morphology

Not specified

products

High levels of L-dopa decarboxylase

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian lung carcinoma

Cell Line Description

Derived from a pleural effusion taken from a 71-year-old Caucasian male suffering from small cell lung carcinoma.

Application

Studies on lung cancer

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. Cells grow as floating aggregates.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service