Skip to Content
Merck
All Photos(1)

Documents

RPMI 2650

88031602, human nose, Epithelial

Synonym(s):

RPMI Cell Culture Medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

RPMI 2650, 88031602

biological source

human nose

growth mode

Adherent

karyotype

2n = 46

morphology

Epithelial

products

Mucoid material

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human nasal septum quasidiploid tumour

Cell Line Description

From the pleural effusion of an extensive malignant tumour of the nasal septum, diagnosed as anaplastic squamous cell carcinoma. Nearly normal karyotype, stability of chromosome number and probable origin from malignant cells make the cell line unique amongst permanent human cell lines. Cells are very small and grow in clusters fusing to form thick layers.

Application

Tumourigenicity studies

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 9,11
D13S317: 11,12
D16S539: 11,12
D5S818: 12,13
D7S820: 8,11
THO1: 6,8
TPOX: 8
vWA: 16,18

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

DNA, RNA, cDNA derived from ECACC mammalian cell lines allow screening for genes or expression patterns to identify lines most suitable for specific research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service