Mycl-9E10
Reactivity: Anti-c-Myc gene product, human, 85102202
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
Mouse x Mouse Hybridoma
ImmunoDon
BALB/c mouse spleen
Myeloma
Sp2/0
Reactivity
Anti-c-Myc gene product, human
technique(s)
cell culture | mammalian: suitable
isotype
IgG1, kappa
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Mouse Sp2/0 x Mouse BALB/c spleen
Cell Line Description
The antibody recognises human p62c-myc gene product and is specific for C-terminal peptide G. The antibody failed to immunoprecipitate p62 c-myc, but recognised the imunoblotted protein. Compared to other hybridomas specific to this antigen (Mycl-8F9, Mycl-3C7, CT9-B7, CT14-B2) Mycl-9E10 is the most sensitive probe for immunoblots. Conformation of antigen is critical for antibody recognition. No reported cross-reaction, with chicken p110gag-myc protein or mouse c-myc protein. This cell line is available as a growing culture only to customers in Europe.
Immunogen
A synthetic peptide, corresponding to residues 408-432(C-terminus of Peptide G) of human p62c-myc sequence, conjugated to KLH
Culture Medium
RPMI 1640 + 2mM glutamine + 10% Foetal Bovine Serum (FBS)
Subculture Routine
Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. Cells are slow to grow. This cell line is available as a growing culture only to customers in Europe.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service