Skip to Content
Merck
All Photos(3)

Key Documents

AV32349

Sigma-Aldrich

Anti-RUVBL1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-ECP54, Anti-NMP238, Anti-Pontin52, Anti-RVB1, Anti-RuvB-like 1 (E. coli), Anti-TIH1, Anti-TIP49, Anti-TIP49A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€498.00

€498.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€498.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€498.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

50 kDa

species reactivity

dog, bovine, mouse, human, horse, rabbit, rat, guinea pig

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... RUVBL1(8607)

General description

Rabbit Anti-RUVBL1 antibody recognizes zebrafish, human, mouse, rat, chicken, canine, and bovine RUVBL1.

Immunogen

Synthetic peptide directed towards the N terminal region of human RUVBL1

Application

Rabbit Anti-RUVBL1 antibody can be used for western blot applications at a concentration of 1μg/ml.

Biochem/physiol Actions

RUVBL1 (RuvB-like AAA ATPase 1) is involved in various cellular processes including transcriptional regulation, DNA replication, DNA damage repair, chromatin remodeling and apoptosis.[1] In the chromatin-remodeling process, it conjugates with other proteins to form the human histone acetylase/chromatin-remodeling complex TIP60, which plays an essential role in transcription and DNA repair. The chromatin-remodeling complex regulates chromatin structure as well as it maintains DNA-based export in the cell. It also functions as a component of the human RNA polymerase II holoenzyme complex, which indicates its role in transcriptional processes. RUVBL1 is also associated with two oncogenic pathways, c-Myc and another, β-catenin pathways. In addition, it also performs in small nucleolar ribonucleoprotein particle assembly, nucleolar localization, and trafficking.[2]

Sequence

Synthetic peptide located within the following region: KIEEVKSTTKTQRIASHSHVKGLGLDESGLAKQAASGLVGQENAREACGV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wolfgang Gartner et al.
Cell motility and the cytoskeleton, 56(2), 79-93 (2003-09-25)
RUVBL1/TIP49a/Pontin52 is a recently identified multi-functional protein with 2 ATP binding (WALKER) sites, which is essential for cell proliferation. We recovered and identified RUVBL1/TIP49a as a tubulin-binding protein from Triton X-100 lysates of U937 promonocytic cells by protein affinity chromatography
X B Qiu et al.
The Journal of biological chemistry, 273(43), 27786-27793 (1998-10-17)
A human protein (RUVBL1), consisting of 456 amino acids (50 kDa) and highly homologous to RuvB, was identified by using the 14-kDa subunit of replication protein A (hsRPA3) as bait in a yeast two-hybrid system. RuvB is a bacterial protein
Pedro M Matias et al.
The Journal of biological chemistry, 281(50), 38918-38929 (2006-10-25)
RuvBL1 is an evolutionarily highly conserved eukaryotic protein belonging to the AAA(+)-family of ATPases (ATPase associated with diverse cellular activities). It plays important roles in essential signaling pathways such as the c-Myc and Wnt pathways in chromatin remodeling, transcriptional and
Jirina Tyleckova et al.
International journal of molecular sciences, 13(12), 15536-15564 (2013-02-28)
A comprehensive proteome map of T-lymphoblastic leukemia cells and its alterations after daunorubicin, doxorubicin and mitoxantrone treatments was monitored and evaluated either by paired comparison with relevant untreated control and using multivariate classification of treated and untreated samples. With the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service