Skip to Content
Merck
All Photos(1)

Key Documents

A8505

Supelco

Androstanolone 17-benzoate

Synonym(s):

17β-Hydroxy-5α-androstan-3-one 17-benzoate, 5α-Androstan-17β-ol-3-one benzoate, Dihydrotestosterone benzoate, Stanolone benzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H34O3
CAS Number:
Molecular Weight:
394.55
Beilstein:
3122672
EC Number:
MDL number:
UNSPSC Code:
41116107

drug control

USDEA Schedule III; Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

SMILES string

C[C@]12CCC(=O)C[C@@H]1CC[C@H]3[C@@H]4CC[C@H](OC(=O)c5ccccc5)[C@@]4(C)CC[C@H]23

InChI

1S/C26H34O3/c1-25-14-12-19(27)16-18(25)8-9-20-21-10-11-23(26(21,2)15-13-22(20)25)29-24(28)17-6-4-3-5-7-17/h3-7,18,20-23H,8-16H2,1-2H3/t18-,20-,21-,22-,23-,25-,26-/m0/s1

InChI key

ZGDZDAPCWHIIKB-LVYWIKMTSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A E Johnson et al.
Endocrinology, 128(2), 891-896 (1991-02-01)
Oxytocin (OT) receptor binding in the ventromedial hypothalamic nucleus is regulated by testosterone (T) in male rats. However, T is metabolized in the brain, and many of the central effects of T are mediated by its metabolites. The experiments reported
F Garza et al.
Journal of animal science, 61(1), 240-244 (1985-07-01)
Eight long-term ovariectomized pony mares were treated with either dihydrotestosterone (DHT) benzoate (400 micrograms/kg body weight) in safflower oil or an equivalent amount of oil every other day for 21 d to determine the effects of DHT on follicle stimulating
D L Thompson et al.
Biology of reproduction, 29(4), 970-976 (1983-11-01)
Sixteen intact cyclic mares were treated on the fourth day of estrus and then every other day for a total of six injections with 1) testosterone propionate, 2) dihydrotestosterone (DHT) benzoate, 3) estradiol (E2) benzoate or 4) safflower oil. Mares
Susan K Putnam et al.
Hormones and behavior, 44(5), 419-426 (2003-12-04)
The medial preoptic area (MPOA) is an important integrative site for male sexual behavior. Dopamine (DA) is released in the MPOA of male rats shortly before and during copulation. The recent presence of testosterone (T) may be necessary for this
Danielle M Cusmano et al.
Endocrinology, 155(1), 204-214 (2013-11-06)
The paucity of clinical and preclinical studies investigating sex differences in sleep has resulted in mixed findings as to the exact nature of these differences. Although gonadal steroids are known to modulate sleep in females, less is known about males.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service