Skip to Content
Merck
All Photos(1)

Documents

A5220

Sigma-Aldrich

N-Acetyl-Val-Glu-Ile-Asp-p-nitroanilide

≥97% (HPLC), powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

form

powder

solubility

DMSO: 2 mM

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](CCC(O)=O)NC(=O)[C@@H](NC(C)=O)C(C)C)C(=O)N[C@@H](CC(O)=O)C(=O)Nc1ccc(cc1)[N+]([O-])=O

InChI

1S/C28H40N6O11/c1-6-15(4)24(33-25(40)19(11-12-21(36)37)31-27(42)23(14(2)3)29-16(5)35)28(43)32-20(13-22(38)39)26(41)30-17-7-9-18(10-8-17)34(44)45/h7-10,14-15,19-20,23-24H,6,11-13H2,1-5H3,(H,29,35)(H,30,41)(H,31,42)(H,32,43)(H,33,40)(H,36,37)(H,38,39)/t15-,19-,20-,23-,24-/m0/s1

InChI key

VXTVKOWKXXPPNE-PZQVQNRFSA-N

Gene Information

human ... CASP6(839)

Amino Acid Sequence

NAc-Val-Glu-Ile-Asp-pNA

Substrates

Colormetric substrate for caspase 6.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gustavo Lemos Rocha et al.
Plant science : an international journal of experimental plant biology, 288, 110232-110232 (2019-09-16)
Seed coat is the tissue which establishes an interface between the seed inner tissues and external environment. Our group has shown that cowpea seed coat undergoes coordinated events of programmed cell death (PCD) during development. In relation to germinating seed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service