Skip to Content
Merck
All Photos(1)

Key Documents

Z640247

Eppendorf® epT.I.P.S. box

volume range 50-1000 μL

Synonym(s):

pipetting tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.01

material

blue rack

sterility

non-sterile

feature

barrier: no

packaging

pack of 96 (includes 1 reusable box)

manufacturer/tradename

Eppendorf® 0030 073.100
Eppendorf® 022491351

L

71 mm

maximum volume

1 mL

volume range

50-1000 μL

Looking for similar products? Visit Product Comparison Guide

General description

Part of the ep T.I.P.S. box and reload system for contamination-free handling of refill trays, with plastic lids protecting the trays from contact during transfer to the working box.
Autoclavable working box, optimized for multichannel pipettes, with removable lid and larger footprint to avoid tipping. Stackable. Supplied full of tips.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Optimized separation of Oligo Standard 6, an internally created HPLC-UV system suitability mix, on Chromolith® RP-18e columns with an evaluation of the effect of flow rates up to 3 mL/min and ion-pairing reagent, TEAA, on the separation.

PhotoKitAlysis Reaction Kit aids in cross-coupling reactions, offering pre-weighed components for optimization.

Characterize mAb monomers, aggregates, and fragments using SEC-UV workflow with Zenix® and Zenix®-C SEC columns.

ICP-MS analyzes 12 heavy metals in hemp-containing food samples using standard addition calibration for accurate quantification.

See All

Protocols

Explore catalysis methods like Suzuki-Miyaura, Buchwald-Hartwig, Heck, Sonagashira, and more with our Palladium Precatalyst Screening Kits.

Complete workflow for the comprehensive analysis of terpenes in cannabis

KitAlysis™ Cu C-N (Buchwald-Hartwig) cross-coupling high-throughput screening kit. Detailed Set-Up User Guide and a downloadable excel file for calculations.

A complete SEC-UV workflow for quantifying monoclonal antibody fragments, monomers, and aggregates, including procedure for a system suitability test and heat stressing of antibodies.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service