Skip to Content
Merck
All Photos(1)

Key Documents

P0098

Sigma-Aldrich

Pediocin from Pediococcus acidilactici

≥95% (HPLC), buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
51102829
NACRES:
NA.85

Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

buffered aqueous solution

concentration

0.1 mg/mL in 0.1 M sodium acetate pH 5.0

antibiotic activity spectrum

Gram-positive bacteria

Mode of action

cell membrane | interferes

shipped in

dry ice

storage temp.

−20°C

General description

Chemical structure: peptide

Application

Pediocins have molecular masses of about 2,700 to 4,000 Da[1]. They are potential food biopreservatives used to control food-borne pathogens and spoilage[1].

Biochem/physiol Actions

Pediocins are class IIa bacteriocins that are produced by Pediococcus acidilactici. They are cationic peptides with molecular masses of about 2,700 to 4,000 Da, that show strong activity against pathogenic bacteria such as Listeria monocytogenes, Clostridicum perfringes, Enterococcus faecalis, and Staphylococcus aureus.4 This antimicrobial action of pediocins is based on interactions with the cytoplasmic membrane, resulting in pore formation and cell death. These interactions also cause a change in membrane potentials, which leads to a loss of the proton motive force and an efflux of intracellular ions, ultimately causing cell death. They have been shown to inhibit Listeria monocytogenes.

Other Notes

50ug
Keep container tightly closed in a dry and well-ventilated place.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M J Pucci et al.
Applied and environmental microbiology, 54(10), 2349-2353 (1988-10-01)
The bacteriocin produced by Pediococcus acidilactici PAC 1.0, previously designated PA-1 bacteriocin, was found to be inhibitory and bactericidal for Listeria monocytogenes. A dried powder prepared from PAC 1.0 culture supernatant fortified with 10% milk powder was found to contain
A K Bhunia
Applied and environmental microbiology, 60(8), 2692-2696 (1994-08-01)
Monoclonal antibody (MAb) R2-AR against pediocin RS2 was developed. Mice were immunized for 12 weeks with pediocin RS2 conjugated to a polyacrylamide gel. Two hybridoma fusions yielded an MAb that in Western blots (immunoblots) reacted only with pediocins RS2 and
Pamela Oliveira de Souza de Azevedo et al.
Scientific reports, 10(1), 12291-12291 (2020-07-25)
Pediococcus pentosaceus LBM 18 has shown potential as producer of an antibacterial and antifungal bacteriocin-like inhibitory substance (BLIS). BLIS inhibited the growth of spoilage bacteria belonging to Lactobacillus, Enterococcus and Listeria genera with higher activity than Nisaplin used as control.

Articles

Ribosomally synthesized antimicrobial peptides are a promising focus in antibiotic research amidst bacterial resistance and emerging infectious diseases.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service