Skip to Content
Merck
All Photos(1)

Key Documents

MIRUP

Sigma-Aldrich

MystiCq® Universal PCR Primer

Sign Into View Organizational & Contract Pricing

Select a Size

500 REACTIONS
€117.00

€117.00


Estimated to ship on26 May 2025



Select a Size

Change View
500 REACTIONS
€117.00

About This Item

UNSPSC Code:
41106308
NACRES:
NA.55

€117.00


Estimated to ship on26 May 2025


form

liquid

concentration

10 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

The MystiCq Universal PCR primer is an integral part of the MystiCq microRNA qPCR Assay System. It has been designed to work specifically with the MystiCq microRNA qPCR Assay primers and the MystiCq microRNA SYBR Green qPCR ReadyMix on cDNA templates generated by the MystiCq microRNA cDNA Synthesis Mix kit.

Application

MystiCq® Universal PCR Primer has been used for the quantitation of microRNA.[1][2]

Legal Information

MystiCq is a registered trademark of QIAGEN Beverly Inc.

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Coding and non-coding variants in the SHOX2 gene in patients with early-onset atrial fibrillation
Sandra Hoffmann
Basic Research in Cardiology, 111 (2016)
High Glucose-Mediated STAT3 Activation in Endometrial Cancer Is Inhibited by Metformin: Therapeutic Implications for Endometrial Cancer
John J. Wallbillich
PLoS ONE, 12(1), e0170318-e0170318 null
Bijay Dhungel et al.
Scientific reports, 8(1), 15539-15539 (2018-10-21)
In hepatocellular carcinoma (HCC), which usually develops in a cirrhotic liver, treatments preserving normal liver function and viability are vitally important. Here, we utilise the differential expression of miRNAs 122a and 199a between normal hepatocytes and HCC to generate vectors
Justin B Collier et al.
Cellular and molecular life sciences : CMLS, 77(18), 3643-3655 (2019-12-25)
Prior studies have established the important role of extracellular signal-regulated kinase 1/2 (ERK1/2) as a mediator of acute kidney injury (AKI). We demonstrated rapid ERK1/2 activation induced renal dysfunction following ischemia/reperfusion (IR)-induced AKI and downregulated the mitochondrial biogenesis (MB) regulator
Yuhao Zhang et al.
American journal of physiology. Lung cellular and molecular physiology, 316(1), L144-L156 (2018-11-02)
Premature male neonates are at a greater risk of developing bronchopulmonary dysplasia (BPD). The reasons underlying sexually dimorphic outcomes in premature neonates are not known. The role of miRNAs in mediating sex biases in BPD is understudied. Analysis of the

Protocols

Method for purification, reverse transcription and quantitative PCR for MicroRNAs using Mysticq reagents

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service