Skip to Content
Merck
All Photos(1)

Key Documents

C1355

Sigma-Aldrich

Z-Leu-Glu(O-Me)-His-Asp(O-Me) fluoromethyl ketone trifluoroacetate salt hydrate

≥90%

Synonym(s):

Z-LEHD-FMK, Z-LE(OMe)HD(OMe)-FMK

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H43FN6O10 · xC2HF3O2 · yH2O
CAS Number:
Molecular Weight:
690.72 (anhydrous free base basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥90%

form

film

storage condition

desiccated

color

colorless to light brown

solubility

DMSO: 10 mg/mL, clear

shipped in

dry ice

storage temp.

−20°C

SMILES string

COC(=O)CC[C@H](NC(=O)[C@H](CC(C)C)NC(=O)OCc1ccccc1)C(=O)N[C@@H](Cc2c[nH]cn2)C(=O)N[C@@H](CC(=O)OC)C(=O)CF

InChI

1S/C32H43FN6O10/c1-19(2)12-24(39-32(46)49-17-20-8-6-5-7-9-20)30(44)36-22(10-11-27(41)47-3)29(43)38-25(13-21-16-34-18-35-21)31(45)37-23(26(40)15-33)14-28(42)48-4/h5-9,16,18-19,22-25H,10-15,17H2,1-4H3,(H,34,35)(H,36,44)(H,37,45)(H,38,43)(H,39,46)/t22-,23-,24-,25-/m0/s1

InChI key

YXRKBEPGVHOXSV-QORCZRPOSA-N

Application

A cell-permeable inhibitor of caspase-9, which exhibits competitive and irreversible inhibition.

Features and Benefits

This compound is featured on the Caspases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer Simpson et al.
American journal of respiratory and critical care medicine, 201(11), 1358-1371 (2020-02-28)
Rationale: Respiratory syncytial virus (RSV) bronchiolitis causes significant infant mortality. Bronchiolitis is characterized by airway epithelial cell (AEC) death; however, the mode of death remains unknown.Objectives: To determine whether necroptosis contributes to RSV bronchiolitis pathogenesis via HMGB1 (high mobility group
Masahiro Kawatani et al.
Journal of neurophysiology, 126(6), 1959-1977 (2021-11-04)
Barrington's nucleus (Bar), which controls micturition behavior through downstream projections to the spinal cord, contains two types of projection neurons, BarCRH and BarESR1, that have different functions and target different spinal circuitry. Both types of neurons project to the L6-S1
Shuheng Lin et al.
Nature communications, 8, 16074-16074 (2017-07-19)
Notch signalling is a causal determinant of cancer and efforts have been made to develop targeted therapies to inhibit the so-called canonical pathway. Here we describe an unexpected pro-apoptotic role of Notch3 in regulating tumour angiogenesis independently of the Notch
Wei-Lin Hu et al.
Frontiers in cellular and infection microbiology, 7, 471-471 (2017-12-01)
Leptospirosis is a global zoonotic infectious disease caused by pathogenic
Hyo-Jin Kim et al.
Journal of translational medicine, 15(1), 123-123 (2017-06-02)
Shikonin, a natural naphthoquinone pigment purified from Lithospermum erythrorhizon, induces necroptosis in various cancer types, but the mechanisms underlying the anticancer activity of shikonin in lung cancer are not fully understood. This study was designed to clarify whether shikonin causes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service