Skip to Content
Merck
All Photos(1)

Documents

A1466

Sigma-Aldrich

N-Acetyl-Trp-Glu-His-Asp-al

≥80% (HPLC), powder

Synonym(s):

Ac-WEHD-CHO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H33N7O9
CAS Number:
Molecular Weight:
611.60
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥80% (HPLC)

form

powder

color

white

solubility

H2O: 1 mg/mL

storage temp.

−20°C

SMILES string

CC(=O)N[C@@H](Cc1c[nH]c2ccccc12)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](Cc3c[nH]cn3)C(=O)N[C@@H](CC(O)=O)C=O

InChI

1S/C28H33N7O9/c1-15(37)32-22(8-16-11-30-20-5-3-2-4-19(16)20)28(44)34-21(6-7-24(38)39)26(42)35-23(9-17-12-29-14-31-17)27(43)33-18(13-36)10-25(40)41/h2-5,11-14,18,21-23,30H,6-10H2,1H3,(H,29,31)(H,32,37)(H,33,43)(H,34,44)(H,35,42)(H,38,39)(H,40,41)/t18-,21-,22-,23-/m0/s1

InChI key

ZSZYUXBVDPGGGX-QGQQZZQASA-N

Biochem/physiol Actions

Very potent caspase 1 and 5 inhibitor.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daichi Sadatomi et al.
Journal of biochemistry, 161(6), 503-512 (2017-01-18)
The NLRP3 inflammasome plays a critical role in the processing and release of inflammatory cytokines, such as interleukin-1β (IL-1β) and IL-18. Accumulating evidence suggests that mitochondria are common mediators of NLRP3 inflammasome activation induced by a wide range of inflammatory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service