Skip to Content
Merck
All Photos(1)

Key Documents

77470

Sigma-Aldrich

Phenanthrene

purum, for fluorescence, ≥97.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10
CAS Number:
Molecular Weight:
178.23
Beilstein:
1905428
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

grade

for fluorescence
purum

Assay

≥97.0% (HPLC)

bp

340 °C (lit.)

mp

98-100 °C (lit.)
98-100 °C

solubility

dioxane: soluble

density

1.063 g/mL at 25 °C (lit.)

fluorescence

λex 254 nm; λem 378 nm in cyclohexane

SMILES string

c1ccc2c(c1)ccc3ccccc23

InChI

1S/C14H10/c1-3-7-13-11(5-1)9-10-12-6-2-4-8-14(12)13/h1-10H

InChI key

YNPNZTXNASCQKK-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Willian G Birolli et al.
Marine pollution bulletin, 129(2), 525-533 (2017-10-23)
The biodegradation of polycyclic aromatic hydrocarbons (PAHs) by marine-derived fungi was reported in this work. Marine-derived fungi (Trichoderma harzianum CBMAI 1677, Cladosporium sp. CBMAI 1237, Aspergillus sydowii CBMAI 935, Penicillium citrinum CBMAI 1186 and Mucor racemosus CBMAI 847) biodegraded anthracene
Marino Marinković et al.
Environmental science & technology, 46(22), 12679-12686 (2012-11-07)
Cellular stress responses are frequently presumed to be more sensitive than traditional ecotoxicological life cycle end points such as survival and growth. Yet, the focus to reduce test duration and to generate more sensitive end points has caused transcriptomics studies
Martin Holmstrup et al.
Environmental pollution (Barking, Essex : 1987), 184, 370-376 (2013-10-08)
Interaction between effects of hazardous chemicals in the environment and adverse climatic conditions is a problem that receives increased attention in the light of climate change. We studied interactive effects of phenanthrene and drought using a test system in which
Jennifer R McKelvie et al.
Environmental pollution (Barking, Essex : 1987), 175, 75-81 (2013-01-23)
We evaluated the correlation between soil organic carbon (OC) content and metabolic responses of Eisenia fetida earthworms after exposure to phenanthrene (58 ± 3 mg/kg) spiked into seven artificial soils with OC contents ranging from 1 to 27% OC. Principal
Geoffrey Marchal et al.
Environmental pollution (Barking, Essex : 1987), 181, 200-210 (2013-07-23)
Sorption of PAHs to carbonaceous soil amendments reduces their dissolved concentrations, limiting toxicity but also potentially biodegradation. Therefore, the maximum abiotic desorption of freshly sorbed phenanthrene (≤5 mg kg(-1)) was measured in three soils amended with activated carbon (AC), biochar

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service