Skip to Content
Merck
All Photos(1)

Key Documents

74032

Sigma-Aldrich

Nitrotetrazolium Blue chloride

≥98.0% (calc. based on dry substance, T)

Synonym(s):

2,2′-bis(4-Nitrophenyl)-5,5′-diphenyl-3,3′-(3,3′-dimethoxy-4,4′-diphenylene)ditetrazolium chloride, 3,3′-(3,3′-Dimethoxy-4,4′-biphenylene)bis[2-(4-nitrophenyl)-5-phenyl-2H-tetrazolium chloride], p-Nitro-Blue tetrazolium chloride, p-Nitrotetrazolium blue, NBT, Nitro BT

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C40H30N10O6 · 2Cl
CAS Number:
Molecular Weight:
817.64
Beilstein:
4115923
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

Assay

≥98.0% (calc. based on dry substance, T)

form

powder

impurities

≤10% solvent content
≤10% water

solubility

methanol: 50 mg/mL, clear, very strongly yellow

storage temp.

2-8°C

SMILES string

[Cl-].[Cl-].COc1cc(ccc1-[n+]2nc(nn2-c3ccc(cc3)[N+]([O-])=O)-c4ccccc4)-c5ccc(c(OC)c5)-[n+]6nc(nn6-c7ccc(cc7)[N+]([O-])=O)-c8ccccc8

InChI

1S/C40H30N10O6.2ClH/c1-55-37-25-29(13-23-35(37)47-43-39(27-9-5-3-6-10-27)41-45(47)31-15-19-33(20-16-31)49(51)52)30-14-24-36(38(26-30)56-2)48-44-40(28-11-7-4-8-12-28)42-46(48)32-17-21-34(22-18-32)50(53)54;;/h3-26H,1-2H3;2*1H/q+2;;/p-2

InChI key

FSVCQIDHPKZJSO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emanuela Romano et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(19), 6140-6145 (2015-04-29)
Enhancing immune responses with immune-modulatory monoclonal antibodies directed to inhibitory immune receptors is a promising modality in cancer therapy. Clinical efficacy has been demonstrated with antibodies blocking inhibitory immune checkpoints such as cytotoxic T lymphocyte-associated antigen 4 (CTLA-4) or PD-1/PD-L1.
Chihiro Ebihara et al.
Human molecular genetics, 24(15), 4238-4249 (2015-05-03)
Seipin, encoded by BSCL2 gene, is a protein whose physiological functions remain unclear. Mutations of BSCL2 cause the most-severe form of congenital generalized lipodystrophy (CGL). BSCL2 mRNA is highly expressed in the brain and testis in addition to the adipose
Ming-Yuan Tsai et al.
Developmental neurobiology, 75(9), 908-926 (2014-12-23)
Krüppel-like factor 8 (Klf8) is a zinc-finger transcription factor implicated in cell proliferation, and cancer cell survival and invasion; however, little is known about its role in normal embryonic development. Here, we show that Klf8 is required for normal cerebellar
Pablo Gómez-Ochoa et al.
Veterinary immunology and immunopathology, 146(1), 97-99 (2012-03-01)
The nitroblue tetrazolium reduction test (NBT) is an assay based on the activation percentage of neutrophils in peripheral blood, that has been proposed for the follow up of canine leishmaniosis owing to the narrow relationship between the molecules involved in
Shih-Yeh Lin et al.
Vaccine, 33(44), 5966-5973 (2015-05-06)
Enterovirus 71 (EV71) causes the outbreaks of hand-foot-and-mouth disease and results in deaths of hundreds of young children. EV71 virus-like particles (VLPs) are empty capsids consisting of viral structural proteins and can elicit potent immune responses, thus holding promise as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service