Skip to Content
Merck
All Photos(1)

Documents

52419

Sigma-Aldrich

Ene-Reductase

recombinant, expressed in E. coli, ≥1 U/mg

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
1.3.-.-
UNSPSC Code:
12352204

recombinant

expressed in E. coli

form

powder

specific activity

≥1 U/mg

storage temp.

2-8°C

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Unit Definition

1 U of enzyme activity is defined as the amount of the enzyme as lyophilized powder that converts 1 μmol 2-cyclohexen-1one per hour

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong Chen et al.
BMC biotechnology, 12, 89-89 (2012-11-28)
Biliverdin IXα is produced when heme undergoes reductive ring cleavage at the α-methene bridge catalyzed by heme oxygenase. It is subsequently reduced by biliverdin reductase to bilirubin IXα which is a potent endogenous antioxidant. Biliverdin IXα, through interaction with biliverdin
Linda T Hiraki et al.
Genetic epidemiology, 37(1), 92-98 (2012-11-09)
The primary circulating form of vitamin D is 25-hydroxy vitamin D (25(OH)D), a modifiable trait linked with a growing number of chronic diseases. In addition to environmental determinants of 25(OH)D, including dietary sources and skin ultraviolet B (UVB) exposure, twin-
Kuan Hu et al.
The Biochemical journal, 449(1), 79-89 (2012-10-12)
TERs (trans-2-enoyl-CoA reductases; EC 1.3.1.44), which specifically catalyse the reduction of crotonyl-CoA to butyryl-CoA using NADH as cofactor, have recently been applied in the design of robust synthetic pathways to produce butan-1-ol as a biofuel. We report in the present
Caroline E Paul et al.
Organic letters, 15(1), 180-183 (2012-12-22)
A series of synthetic nicotinamide cofactors were synthesized to replace natural nicotinamide cofactors and promote enoate reductase (ER) catalyzed reactions without compromising the activity or stereoselectivity of the bioreduction process. Conversions and enantioselectivities of >99% were obtained for C═C bioreductions
Ayushi Kamthan et al.
Scientific reports, 2, 951-951 (2012-12-12)
Crop genetic engineering mostly aims at improving environmental stress (biotic and abiotic) tolerance as well as nutritional quality. Empowering a single crop with multiple traits is highly demanding and requires manipulation of more than one gene. However, we report improved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service