Skip to Content
Merck

Skip To

27673

Collagenase Substrate

BioReagent, ≥95.0% (HPLC), powder, suitable for quantitative collagenase determination

Synonym(s):

Z-Gly-Pro-Gly-Gly-Pro-Ala-OH

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mg
Check Cart for Availability
€ 376,00
€ 319,60

About This Item

Empirical Formula (Hill Notation):
C27H36N6O9
CAS Number:
Molecular Weight:
588.61
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352204
EC Number:
235-973-2
MDL number:
Beilstein/REAXYS Number:
6039452

€ 319,60

List Price€ 376,00Save 15%
Web-Only Promotion

Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Collagenase Substrate, BioReagent, For quantitative collagenase determination

Quality Segment

product line

BioReagent

assay

≥95.0% (HPLC)

form

powder

shipped in

wet ice

storage temp.

−20°C

SMILES string

C[C@H](NC(=O)[C@@H]1CCCN1C(=O)CNC(=O)CNC(=O)[C@@H]2CCCN2C(=O)CNC(=O)OCc3ccccc3)C(O)=O

InChI

1S/C27H36N6O9/c1-17(26(39)40)31-25(38)20-10-6-12-33(20)22(35)14-28-21(34)13-29-24(37)19-9-5-11-32(19)23(36)15-30-27(41)42-16-18-7-3-2-4-8-18/h2-4,7-8,17,19-20H,5-6,9-16H2,1H3,(H,28,34)(H,29,37)(H,30,41)(H,31,38)(H,39,40)/t17-,19-,20-/m0/s1

InChI key

YMMPZCZOLKBHAP-IHPCNDPISA-N


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
2766727674SML2836
assay

≥95.0% (HPLC)

assay

-

assay

-

assay

≥95% (HPLC)

Quality Level

200

Quality Level

100

Quality Level

100

Quality Level

100

form

powder

form

powder or crystals

form

powder

form

lyophilized powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

-

product line

BioReagent

product line

-

product line

-

product line

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters