Key Documents
Z804479
Hellma® liquid calibration standards
UV35, for testing the wavelength accuracy
About This Item
UNSPSC Code:
12352200
NACRES:
SB.32
manufacturer/tradename
Hellma 667-UV35
technique(s)
UV/Vis spectroscopy: suitable
General description
Certified liquid Hellma® calibration standard to enable the checking of spectrophotometers with respect to wavelength accuracy.
Consisting of :
rare earth (667-UV35, wavelength 201; 211; 222; 239, 252 nm)
Other Notes
The most current version of Hellma's handling guidelines for calibration standards can be accessed on the Hellma Analytics web site.
Legal Information
Hellma is a registered trademark of Hellma GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)