Skip to Content
Merck
All Photos(1)

Key Documents

Y0000227

Meso-rich exametazime

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Exametazime

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H28N4O2
CAS Number:
Molecular Weight:
272.39
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

meso-rich exametazime

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C13H28N4O2/c1-9(11(3)16-18)14-7-13(5,6)8-15-10(2)12(4)17-19/h9-10,14-15,18-19H,7-8H2,1-6H3/b16-11+,17-12+/t9-,10-/m1/s1

InChI key

BPNZYADGDZPRTK-UDUYQYQQSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Meso-rich exametazime EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S J Colloby et al.
Neurobiology of aging, 32(2), 293-301 (2009-03-10)
To investigate differences in distribution of α4β2 subtypes of nicotinic acetylcholine receptors (nAChRs) using the ligand ¹²³I-5-Iodo-3-[2(S)-2-azetidinylmethoxy] pyridine (5IA-85380) and single photon emission computed tomography (SPECT) in subjects with vascular dementia and age-matched controls. ¹²³I-5IA-85380 binding was compared to corresponding
C Solanki et al.
Nuclear medicine communications, 19(6), 567-572 (1999-05-11)
The objective of this study was to increase the stability of 99Tcm-exametazime and to investigate the effects of relaxing the eluate restrictions imposed by the manufacturer. We added 1800 MBq freshly eluted pertechnetate to 0.5 ml aliquots of stannous-enhanced exametazime
Pierre-Yves Litzler et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 51(7), 1044-1048 (2010-06-18)
We report our experience with using integrated molecular and anatomic hybrid imaging to assess infection in patients who have a left-ventricular-assist device (LVAD). Thirteen (99m)Tc-exametazime-leukocyte planar and SPECT/CT scans were obtained for 8 consecutive patients who had an implanted LVAD.
N Nikolić et al.
Journal of pharmaceutical and biomedical analysis, 32(6), 1159-1166 (2003-08-09)
The diastereo-enantio separation of isomeric mixtures of exametazime (HM-PAO) by liquid chromatography is described using an achiral sorbent (RP-18). A chiral eluent with the initial complex of Cu(II) and the optically active selector N,N-dimethyl-l-phenylalanine (l-DM-PhA), based on the ligand-exchange principle
D Vanderghinste et al.
Journal of pharmaceutical and biomedical analysis, 32(4-5), 679-685 (2003-08-06)
99mTc-exametazime (99mTc-d,l-HMPAO, 99mTc-d,l-hexamethylpropyleneamine oxime) is a neutral rather unstable complex of short-lived 99mTc (t(1/2)=6 h) with the d,l-isomer (mixture of D,D- and L,L-isomers) of a bis-amine bis-oxime tetraligand. It is widely used for measurement of regional cerebral perfusion in nuclear

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service