Skip to Content
Merck
All Photos(1)

Documents

SMB00960

Sigma-Aldrich

Etoposide quinone

Synonym(s):

Etoposide 3′,4′-quinone, Etoposide o-quinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H28O13
CAS Number:
Molecular Weight:
572.51
UNSPSC Code:
12352201
NACRES:
NA.79

biological source

synthetic

Quality Level

grade

research grade

Assay

≥93% (H-NMR)

form

solid

technique(s)

HPLC: suitable

color

dark red

mp

200—210 °C

storage temp.

−20°C

SMILES string

Canonical SMILES: CC1OCC2C(O1)C(C(C(O2)OC3C4COC(=O)C4C(C5=CC6=C(C=C35)OCO6)C7=CC(=O)C(=O)C(=C7)OC)O)OIsomeric SMILES: CC1OC[C@@H]2[C@@H](O1)[C@@H]([C@H]([C@@H](O2)OC3C4COC(=O)C4C(C5=CC6=C(C=C35)OCO6)C7=CC(=O)C(=O)C(=C7)OC)O)O

InChI

1S/C28H28O13/c1-10-35-8-19-26(39-10)23(31)24(32)28(40-19)41-25-13-6-17-16(37-9-38-17)5-12(13)20(21-14(25)7-36-27(21)33)11-3-15(29)22(30)18(4-11)34-2/h3-6,10,14,19-21,23-26,28,31-32H,7-9H2,1-2H3/t10?,14?,19-,20?,21?,23-,24-,25?,26-,28+/m1/s1

InChI key

SBLYXIKLMHGUJZ-LBMXFXLMSA-N

General description

Etoposide quinone, a major metabolite of the anticancer drug etoposide, is believed to play a role in the development of secondary acute myeloid leukemias (t-AMLs) in some etoposide patients. It acts as a covalent poison of human topoisomerase IIβ, inducing approximately four times more topoisomerase IIβ-mediated DNA cleavage than its parent drug, etoposide. This metabolite is also a potent irreversible inhibitor of TCPTP protein tyrosine phosphatase, with an IC50 of 7 μM. Its inhibition of TCPTP leads to an increase in STAT1 tyrosine phosphorylation, contributing to the leukemogenic effects associated with etoposide. Understanding the unique properties and mechanisms of etoposide quinone is essential for unraveling its potential impact on cancer research, specifically its association with the development of t-AMLs.

Application

Etoposide quinone can be used in biochemical, metabolomics and cancer research

Biochem/physiol Actions

Etoposide undergoes a metabolic transformation by CYP3A4, resulting in the formation of etoposide catechol, which can be subsequently oxidized to produce etoposide quinone.

Other Notes

For additional information on our range of Biochemicals, please complete this form.
To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicholas A Smith et al.
Biochemistry, 53(19), 3229-3236 (2014-04-29)
Etoposide is a topoisomerase II poison that is utilized to treat a broad spectrum of human cancers. Despite its wide clinical use, 2-3% of patients treated with etoposide eventually develop treatment-related acute myeloid leukemias (t-AMLs) characterized by rearrangements of the
Elizabeth G Gibson et al.
Chemical research in toxicology, 29(9), 1541-1548 (2016-08-18)
Topoisomerase II is an essential nuclear enzyme involved in regulating DNA topology to facilitate replication and cell division. Disruption of topoisomerase II function by chemotherapeutic agents is in use as an effective strategy to fight cancer. Etoposide is an anticancer
Qing Nian et al.
Molecular pharmacology, 96(2), 297-306 (2019-06-22)
Etoposide is a widely prescribed anticancer drug that is, however, associated with an increased risk of secondary leukemia. Although the molecular basis underlying the development of these leukemias remains poorly understood, increasing evidence implicates the interaction of etoposide metabolites [i.e.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service