Skip to Content
Merck
All Photos(1)

Documents

PHR1625

Supelco

Rufinamide

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Rufinamide, 1-[(2,6-difluorophenyl)methyl]-1H-1,2,3-Triazole-4-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8F2N4O
CAS Number:
Molecular Weight:
238.19
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1606401

API family

rufinamide

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

NC(=O)c1cn(Cc2c(F)cccc2F)nn1

InChI

1S/C10H8F2N4O/c11-7-2-1-3-8(12)6(7)4-16-5-9(10(13)17)14-15-16/h1-3,5H,4H2,(H2,13,17)

InChI key

POGQSBRIGCQNEG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Rufinamide is an antiepileptic drug, used as an adjunctive for the treatment of seizures associated with Lennox−Gastaut syndrome. Its mode of action involves prolonging the refractory period of voltage-dependent sodium channels via stabilization of the sodium channel inactive state, thus effectively keeping the ion channels closed.

Application

Rufinamide may be used as a pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations using reversed-phase high-performance liquid chromatography technique.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Broad-spectrum anticonvulsant.

Diagnostic Implications

Effective therapy for partial seizures in adults and childhood seizures in Lennox-Gastaut syndrome.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA5294 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stability indicating liquid chromatographic method for the quantitative determination of rufinamide in pharmaceutical dosage forms
Annapurna MM, et al.
Journal of Drug Delivery and Therapeutics, 2(4), 66-70 (2012)
Development and validation of a stability indicating RP-HPLC method for the determination of Rufinamide
Kumar PSB, et al.
Journal of Pharmaceutical Analysis, 3(1), 66-70 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service