Skip to Content
Merck
All Photos(1)

Documents

G7012

Supelco

Guaiacol glyceryl ether carbamate

analytical standard

Synonym(s):

Methocarbamol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15NO5
CAS Number:
Molecular Weight:
241.24
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

NC(OCC(O)COC1=C(OC)C=CC=C1)=O

InChI

1S/C11H15NO5/c1-15-9-4-2-3-5-10(9)16-6-8(13)7-17-11(12)14/h2-5,8,13H,6-7H2,1H3,(H2,12,14)

InChI key

GNXFOGHNGIVQEH-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wuyi Zha et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 878(9-10), 831-835 (2010-02-23)
A simple and reproducible high performance liquid chromatography-tandem mass spectrometric method was developed for methocarbamol analysis in human plasma. Methocarbamol and the internal standard (IS) were extracted by a protein precipitation method. Under isocratic separation condition the chromatographic run time
Maryam Sahebari et al.
Acta medica Iranica, 49(3), 192-194 (2011-06-18)
Descriptive case report of a 42-year old woman with coetaneous vasculitis, and severe abdominal pain, which was led to diagnostic laparotomy. These presentations are probably as a side effect of Methocarbamol injection. This is the first report according to our
50 Years Ago in CORR: A clinical study of 46 males with low-back disorders treated with Methocarbamol. Andres Grisolia MD and J.E.M. Thomson CORR 1959;13:299-304.
Richard A Brand
Clinical orthopaedics and related research, 467(10), 2752-2754 (2009-07-29)
Glen Jickling et al.
Epileptic disorders : international epilepsy journal with videotape, 11(4), 329-332 (2009-12-10)
Acetaminophen is a widely used analgesic that can cause acute liver failure when consumed above a maximum daily dose. Certain patients may be at increased risk of hepatocellular damage even at conventional therapeutic doses. We report a case of a
Mohammad Bagher Gholivand et al.
Talanta, 85(3), 1680-1688 (2011-08-03)
Molecularly imprinted polymers (MIPs) with high selectivity toward methocarbamol have been computationally designed and synthesized based on the general non-covalent molecular imprinting approach. A virtual library consisting of 18 functional monomers was built and possible interactions between the template and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service