Skip to Content
Merck
All Photos(1)

Documents

75412

Supelco

Sennoside B

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H38O20
CAS Number:
Molecular Weight:
862.74
Beilstein:
5723747
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥90.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤10.0% water

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@@H](Oc2cccc3[C@@H]([C@H]4c5cccc(O[C@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O)c5C(=O)c7c(O)cc(cc47)C(O)=O)c8cc(cc(O)c8C(=O)c23)C(O)=O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C42H38O20/c43-11-23-31(47)35(51)37(53)41(61-23)59-21-5-1-3-15-25(17-7-13(39(55)56)9-19(45)27(17)33(49)29(15)21)26-16-4-2-6-22(60-42-38(54)36(52)32(48)24(12-44)62-42)30(16)34(50)28-18(26)8-14(40(57)58)10-20(28)46/h1-10,23-26,31-32,35-38,41-48,51-54H,11-12H2,(H,55,56)(H,57,58)/t23-,24-,25-,26+,31-,32-,35+,36+,37-,38-,41-,42?/m1/s1

InChI key

IPQVTOJGNYVQEO-OVYIKYHGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sennoside B is a plant-derived anthranoid and is one of the active constituents of the senna drug. It is primarily used as a laxative and cathartic for treating constipation and bowel evacuation.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Sennoside B may be used as a reference standard for the determination of sennoside B in pharmaceutical formulations by high-performance liquid chromatography method.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Stimulant laxatives and opioid-induced constipation.
Twycross R, et al.
Journal of Pain and Symptom Management, 43(2), 306-313 (2012)
Validated HPLC method for determination of sennosides A and B in senna tablets.
Sun SW and Su HT
Journal of Pharmaceutical and Biomedical Analysis, 29(5), 881-894 (2002)
Manuk N Manukyan et al.
Surgical laparoscopy, endoscopy & percutaneous techniques, 21(2), 90-93 (2011-04-08)
The aim of this study is to assess 2 different bowel-cleansing agents. The patients were prospectively randomized to 2 arms of sodium phosphate versus Sennoside A+B calcium preparation. Laboratory assessment, body weight, height, and vital signs were obtained at baseline
Constipation and aspiration pneumonia.
Maki Takahashi et al.
Geriatrics & gerontology international, 12(3), 570-571 (2012-06-21)
Acute hemorrhagic colitis following administration of sennosides for colon cleansing.
Burhan Ozdil et al.
The Annals of pharmacotherapy, 44(4), 770-771 (2010-03-25)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service