Skip to Content
Merck
All Photos(2)

Documents

45965

Supelco

1,2-Dinitrobenzene

analytical standard

Synonym(s):

1,2-DNB

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(NO2)2
CAS Number:
Molecular Weight:
168.11
Beilstein:
642224
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

319 °C/773 mmHg (lit.)

mp

114-117 °C (lit.)

application(s)

environmental

format

neat

SMILES string

[O-][N+](=O)c1ccccc1[N+]([O-])=O

InChI

1S/C6H4N2O4/c9-7(10)5-3-1-2-4-6(5)8(11)12/h1-4H

InChI key

IZUKQUVSCNEFMJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2-Dinitrobenzene belongs to the class of nitroaromatic compounds, which are utilized for the manufacture of pesticides, explosives, polymers, etc. They can pose a potential risk to the environment because of their toxic nature.

Application

1,2-Dinitrobenzene has been used as an analytical standard for the determination of the analyte in organic explosives investigated during the forensic analysis by liquid chromatography-atmospheric pressure chemical ionization mass spectrometry (LC-APCI-MS). It may be used as an analytical standard for the determination of the analyte in aqueous samples, environmental samples and human biological samples by various chromatographic techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Application of DLLME based on the solidification of floating organic droplets for the determination of dinitrobenzenes in aqueous samples
Wu Y, et al.
Chromatographia, 72(7-8), 695-699 (2010)
J V Barbo et al.
Journal of immunology (Baltimore, Md. : 1950), 154(4), 1551-1559 (1995-02-15)
A population of CD8+ T cells from dinitrobenzene sulfonate-primed mice produce soluble effector molecules that down-regulate the magnitude of dinitrophenol-specific contact hypersensitivity reactions. These soluble molecules express the binding specificity and serologic determinants of alpha/beta TCR. To examine the requirement
H Tamaki et al.
Journal of bacteriology, 181(9), 2958-2962 (1999-04-28)
Glutathione S-transferases (GSTs) Y-1 and Y-2 from the yeast Issatchenkia orientalis were purified by passage through a glutathione-agarose column, and the cDNA for GST Y-1 was cloned and sequenced. The deduced amino acid sequence consisted of 188 residues with a
P A Cossum et al.
Drug metabolism and disposition: the biological fate of chemicals, 13(6), 664-668 (1985-11-01)
The metabolism of radiolabeled dinitrobenzene (DNB) isomers was compared in hepatocytes and hepatic subcellular fractions isolated from male Fischer-344 rats. Under aerobic conditions, reduction was the major metabolic pathway for m- and p- DNB in hepatocytes with m- and p-nitroaniline
Tomoko Andoh et al.
FEBS letters, 525(1-3), 131-134 (2002-08-07)
In Saccharomyces cerevisiae, the overexpression of ROD1 confers resistance to o-dinitrobenzene (o-DNB), a representative of target drugs of glutathione S-transferase. The roles of Rod1 in drug resistance have remained to be determined. We isolated the rog3 mutation as a suppressor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service