Skip to Content
Merck
All Photos(1)

Documents

34111

Supelco

Eprinomectin solution

100 μg/mL in acetonitrile, PESTANAL®, analytical standard

Synonym(s):

(4′’R)-4″-(Acetylamino)-4″-deoxy-avermectin B1 solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

single component solution

storage temp.

2-8°C

InChI

1S/C49H73NO14/c1-25(2)43-28(5)17-18-48(64-43)23-35-20-34(63-48)16-15-27(4)44(26(3)13-12-14-33-24-57-46-42(52)29(6)19-36(47(53)60-35)49(33,46)54)61-40-22-38(56-11)45(31(8)59-40)62-39-21-37(55-10)41(30(7)58-39)50-32(9)51/h12-15,17-19,25-26,28,30-31,34-46,52,54H,16,20-24H2,1-11H3,(H,50,51)/b13-12-,27-15-,33-14-/t26-,28-,30-,31-,34+,35-,36-,37+,38-,39-,40-,41+,42+,43+,44-,45+,46+,48+,49+/m0/s1

InChI key

WPNHOHPRXXCPRA-TVXIRPTOSA-N

General description

Eprinomectin is a veterinary drug, belonging to avermectins, a class of macrocyclic lactones. It is widely used as an antiparasitic agent for farm animals and shows a broad spectrum of activity against parasites such as nematodes, insects and mites.

Application

Eprinomectin solution may be used as an analytical reference standard for estimating the analyte in bovine urine using high-performance liquid chromatography with fluorescence detection.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of eprinomectin in bovine urine and feces using HPLC with fluorescence detection.
Jiang H, et al.
Chromatographia, 66(5-6), 411-414 (2007)
Application of QuEChERS-High Performance Liquid Chromatography with Postcolumn Fluorescence Derivatization (HPLC-FLD) method to analyze Eprinomectin B1a residues from a pour-on conditioning in bovine edible tissues.
Csuma A, et al.
open chemistry, 13(1), 411-414 (2015)
S Rehbein et al.
Veterinary parasitology, 192(4), 353-358 (2013-01-01)
The efficacy of eprinomectin in an extended-release injection (ERI) formulation was determined in cattle harboring naturally acquired infestations of first- or second- and third-stage larvae of Hypoderma spp. in three studies conducted according to the same protocol in the USA
M D Soll et al.
Veterinary parasitology, 192(4), 313-320 (2013-01-01)
A series of 10 dose confirmation studies was conducted to evaluate the persistent activity of an extended-release injectable (ERI) formulation of eprinomectin against single point challenge infections of gastrointestinal and pulmonary nematodes of cattle. The formulation, selected based on the
Cengiz Gokbulut et al.
American journal of veterinary research, 72(12), 1639-1645 (2011-12-01)
To investigate plasma disposition, concentration in the hair, and anthelmintic efficacy of eprinomectin after topical administration in donkeys. 12 donkeys naturally infected with strongyle nematodes. The pour-on formulation of eprinomectin approved for use in cattle was administered topically to donkeys

Protocols

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service