31744
2-Thiouracil
VETRANAL®, analytical standard
Synonym(s):
4-Hydroxy-2-mercaptopyrimidine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
grade
analytical standard
product line
VETRANAL®
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
mp
>300 °C (lit.)
application(s)
forensics and toxicology
pharmaceutical (small molecule)
veterinary
format
neat
SMILES string
O=C1NC(=S)NC=C1
InChI
1S/C4H4N2OS/c7-3-1-2-5-4(8)6-3/h1-2H,(H2,5,6,7,8)
InChI key
ZEMGGZBWXRYJHK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Legal Information
VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Chemical communications (Cambridge, England), 47(41), 11498-11500 (2011-09-22)
A nonenzymatic method for the selective detection and quantification of serum uric acid (UA) using 2-thiouracil (2-TU) tailored Au nanoparticles is developed. The H-bonding interaction of UA with functionalized Au nanoparticles brings instantaneous visible color change and paves the way
The journal of physical chemistry. A, 116(44), 10712-10720 (2012-10-16)
The reaction of hydroxyl radical ((•)OH) with the nucleic acid base analogue 2-thiouracil (1) has been studied by pulse radiolysis experiments and DFT. The generic intermediate radicals feasible for the (•)OH reactions with 1, namely, one electron oxidation product (1(•+))
Journal of agricultural and food chemistry, 60(32), 7769-7776 (2012-07-28)
In recent years, the frequent detection of the banned substance thiouracil (TU) in livestock urine has been related to its endogenous formation following consumption of glucosinolate-rich Brassicaceae crops. Besides, TU residues have been recovered in these crops upon plant-derived myrosinase
Molecules (Basel, Switzerland), 17(8), 9868-9886 (2012-08-21)
Several novel 6-aryl-5-cyano thiouracil derivatives were synthesized and explored for their activities as antibacterial, antifungal and anticancer agents. The antimicrobial evaluation revealed that compounds 7b and 7c possessed superior antibacterial activity against the Gram positive bacteria S. aureus and B.
Bioorganic & medicinal chemistry, 19(7), 2443-2449 (2011-03-15)
4-Pyrimidinone ribofuranoside (H(2)o(4)U) and 4-pyrimidinone 2'-deoxyribofuranoside (dH(2)o(4)U) were synthesized by the oxidative desulfurization of parent 2-thiouracil nucleosides with m-chloroperbenzoic acid. The crystal structures of H(2)o(4)U and dH(2)o(4)U and their conformations in solution were determined and compared with corresponding 2-thiouracil and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service