Skip to Content
Merck
All Photos(1)

Key Documents

23510

Sigma-Aldrich

2-Chloro-1,3,2-benzodioxaborole

purum, ≥97.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4BClO2
CAS Number:
Molecular Weight:
154.36
Beilstein:
2093985
MDL number:
UNSPSC Code:
12352101

grade

purum

Assay

≥97.0% (AT)

mp

56-58 °C (lit.)
56-58 °C

storage temp.

2-8°C

SMILES string

Clb1oc2ccccc2o1

InChI

1S/C6H4BClO2/c8-7-9-5-3-1-2-4-6(5)10-7/h1-4H

InChI key

AZYGEWXDKHFOKB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reagent for the selective cleavage of many protecting groups (e.g. MEM-ethers, Z-amines, BOC-amines etc); Transformation of urethanes into isocyanates

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V.L.K. Valli et al.
The Journal of Organic Chemistry, 60, 257-257 (1995)
R.K. Boeckmann et al.
Tetrahedron Letters, 26, 1411-1411 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service