Skip to Content
Merck
All Photos(1)

Key Documents

21052

Sigma-Aldrich

Calcium acetate hydrate

purum p.a., ≥90% (KT), powder or chunks

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COO)2Ca · xH2O
CAS Number:
Molecular Weight:
158.17 (anhydrous basis)
Beilstein:
3692527
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

purum p.a.

Assay

≥90% (KT)

form

powder or chunks

loss

≤10% loss on drying, 200 °C

pH

7.0-8.0 (25 °C, 50 mg/mL in H2O)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤500 mg/kg

cation traces

Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤200 mg/kg
Na: ≤200 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

O.CC(=O)O[Ca]OC(C)=O

InChI

1S/2C2H4O2.Ca.H2O/c2*1-2(3)4;;/h2*1H3,(H,3,4);;1H2/q;;+2;/p-2

InChI key

XQKKWWCELHKGKB-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenqiang Liu et al.
Environmental science & technology, 44(2), 841-847 (2009-12-25)
A screening of potential calcium precursors for the production of CaO sorbents for CO(2) capture at high temperature was conducted in this work. The precursors studied include microsized calcium carbonate (CC-CaO), calcium hydroxide (CH-CaO), nanosized (<70 nm) calcium carbonate (CC70
Anthony W Musumeci et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 67(3-4), 649-661 (2006-10-31)
A comprehensive spectroscopic analysis consisting of Raman, infrared (IR) and near-infrared (NIR) spectroscopy was undertaken on two forms of calcium acetate with differing degrees of hydration. Monohydrate (Ca(CH(3)COO)(2).H(2)O) and half-hydrate (Ca(CH(3)COO)(2).0.5H(2)O) species were analysed. Assignments of vibrational bands due to
CMR La 0.7 Ca 0.3 MnO 3 and La 0.7 Sr 0.3 MnO 3 thin films fabricated by sol-gel method.
Cheng ZX, et al.
Journal of Crystal Growth, 275(1), e2415-e2419 (2005)
Dilek Teker et al.
Materials science & engineering. C, Materials for biological applications, 48, 579-585 (2015-01-13)
An innovative multi-layer coating comprising a bioactive compound layer (consisting of hydroxyapatite and calcium titanate) with an underlying titanium oxide layer (in the form of anatase and rutile) has been developed on Grade 4 quality commercially pure titanium via a
Hamid Esfahani et al.
Journal of colloid and interface science, 443, 143-152 (2014-12-30)
Surface modification of electrospun polymeric membrane surfaces is a critical step towards the separation process including protein adsorption. In this study, the electrospun Nylon fibers was incorporated with positively charged zinc doped hydroxyapatite (HAp) nanoparticles to study the adsorption of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service