Skip to Content
Merck
All Photos(1)

Key Documents

19880

Sigma-Aldrich

N-Butyldiethanolamine

purum, ≥99.0% (GC)

Synonym(s):

2,2′-Butyliminodiethanol, N,N-Bis(2-hydroxyethyl)butylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3N(CH2CH2OH)2
CAS Number:
Molecular Weight:
161.24
Beilstein:
1739642
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

5.5 (vs air)

vapor pressure

1 mmHg ( 25 °C)

grade

purum

Assay

≥99.0% (GC)

impurities

≤0.3% water

refractive index

n20/D 1.463 (lit.)
n20/D 1.463

bp

273-275 °C/741 mmHg (lit.)

mp

−70 °C (lit.)

density

0.986 g/mL at 25 °C (lit.)

SMILES string

CCCCN(CCO)CCO

InChI

1S/C8H19NO2/c1-2-3-4-9(5-7-10)6-8-11/h10-11H,2-8H2,1H3

InChI key

GVNHOISKXMSMPX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-Butyldiethanolamine was used in the synthesis of 4,4′-diphenylmethane diisocyanate (MDI).

replaced by

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

265.1 °F - closed cup

Flash Point(C)

129.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anning Wang et al.
International journal of pharmaceutics, 441(1-2), 30-39 (2012-12-25)
A series of temperature- and pH-responsive polyurethanes based on hexamethylene diisocyanate (HDI) and 4,4'-diphenylmethane diisocyanate (MDI) were synthesized by a coupling reaction with bis-1,4-(hydroxyethyl) piperazine (HEP), N-methyldiethanolamine (MDEA) and N-butyldiethanolamine (BDEA), respectively. The chemical structure, molecular weight, thermal property and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service