Skip to Content
Merck
All Photos(1)

Key Documents

192503

Sigma-Aldrich

10-Methyl-9(10H)-acridone

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11NO
CAS Number:
Molecular Weight:
209.24
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

Assay

99%

mp

204-207 °C (lit.)

SMILES string

CN1c2ccccc2C(=O)c3ccccc13

InChI

1S/C14H11NO/c1-15-12-8-4-2-6-10(12)14(16)11-7-3-5-9-13(11)15/h2-9H,1H3

InChI key

XUVKSPPGPPFPQN-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Devika Sasikumar et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 26(9), 2060-2066 (2019-12-15)
The conversion of solar energy to thermal, chemical, or electrical energy attracts great attention in chemistry and physics. There has been a considerable effort for the efficient extraction of photons throughout the entire solar spectrum. In this work light energy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service