Skip to Content
Merck
All Photos(1)

Key Documents

18226

Supelco

1,2-Dilinoleoyl-3-oleoyl-rac-glycerol

analytical standard

Synonym(s):

1,2-Di(cis-9,12-octadecadienoyl)-3-(cis-9-octadecenoyl)-rac-glycerol, OLL

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C57H100O6
CAS Number:
Molecular Weight:
881.40
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

form

viscous liquid

shelf life

limited shelf life, expiry date on the label

application(s)

food and beverages

format

neat

functional group

ester
ketone

storage temp.

−20°C

SMILES string

O=C(OC(COC(CCCCCCC/C=C\C/C=C\CCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O)CCCCCCC/C=C\C/C=C\CCCCC

InChI

1S/C57H100O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-49-55(58)61-52-54(63-57(60)51-48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)53-62-56(59)50-47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h16,18-19,21,25-30,54H,4-15,17,20,22-24,31-53H2,1-3H3/b19-16-,21-18-,28-25-,29-26-,30-27-

InChI key

VVEBTVMJPTZDHO-WECKWCTPSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

remarks on GC:
~33% oleic acid
~66% linoleic acid

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service