14495
L-Cysteine 0.1 M solution
Synonym(s):
L-Cysteine solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
storage temp.
2-8°C
InChI
1S/C3H7NO2S/c4-2(1-7)3(5)6/h2,7H,1,4H2,(H,5,6)
InChI key
XUJNEKJLAYXESH-UHFFFAOYSA-N
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Nitric oxide quenches the fire in heart mitochondria.
Nature medicine, 19(6), 666-667 (2013-06-08)
Proceedings of the National Academy of Sciences of the United States of America, 110(25), 10153-10158 (2013-05-31)
Characterization of the mature protein complement in cells is crucial for a better understanding of cellular processes on a systems-wide scale. Toward this end, we used single-dimension ultra-high-pressure liquid chromatography mass spectrometry to investigate the comprehensive "intact" proteome of the
Nature medicine, 19(6), 753-759 (2013-05-28)
Oxidative damage from elevated production of reactive oxygen species (ROS) contributes to ischemia-reperfusion injury in myocardial infarction and stroke. The mechanism by which the increase in ROS occurs is not known, and it is unclear how this increase can be
Cell, 153(6), 1379-1393 (2013-06-12)
Some species mount a robust antibody response despite having limited genome-encoded combinatorial diversity potential. Cows are unusual in having exceptionally long CDR H3 loops and few V regions, but the mechanism for creating diversity is not understood. Deep sequencing reveals
Fiziolohichnyi zhurnal (Kiev, Ukraine : 1994), 59(1), 40-46 (2013-05-30)
We studied parameters of gastric secretion in pylorus-ligated rat and blood flow in the rat gastric mucosa under the influence of drug corvitin used intragastrically in doses of 2.5 and 5 mg/kg. Biochemical analysis of gastric juice was based on
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service