Skip to Content
Merck
All Photos(1)

Key Documents

03890585

Taxifolin

primary reference standard

Synonym(s):

(2R,3R)-3,3′,4′,5,7-Pentahydroxyflavanone, (2R,3R)-Dihydroquercetin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12O7
CAS Number:
Molecular Weight:
304.25
Beilstein:
5299277
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

application(s)

food and beverages

storage temp.

−20°C

SMILES string

O[C@@H]1[C@H](Oc2cc(O)cc(O)c2C1=O)c3ccc(O)c(O)c3

InChI

1S/C15H12O7/c16-7-4-10(19)12-11(5-7)22-15(14(21)13(12)20)6-1-2-8(17)9(18)3-6/h1-5,14-19,21H/t14-,15+/m0/s1

InChI key

CXQWRCVTCMQVQX-LSDHHAIUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference Standard in the analysis of herbal medicinal products

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sharad Verma et al.
Journal of molecular graphics & modelling, 37, 27-38 (2012-05-23)
Hsp90 (heat shock protein 90), a molecular chaperone, stabilizes more than 200 mutated and over expressed oncogenic proteins in cancer development. Cdc37 (cell division cycle protein 37), a co-chaperone of Hsp90, has been found to facilitate the maturation of protein
Jan Vacek et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 899, 109-115 (2012-06-06)
Liquid chromatography (LC) on various stationary phases was used for the metabolite profile analysis of quercetin, rutin, isoquercitrin and taxifolin. The metabolites were obtained using an in vitro model system of human and rat hepatocytes in the form of cell
Yuzhi Rong et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 93, 235-239 (2012-04-10)
The antioxidant capacities of the selected flavonoids quercetin, luteolin and taxifolin have been investigated at density functional level of theory with the aim of verifying the cellular antioxidant activity (CAA) values representative of experimental findings. The selected flavonoids were believed
Mathias A E Frevel et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 50(12), 4316-4324 (2012-09-18)
Enzogenol® pine bark extract is a dietary supplement and food ingredient produced by water extraction of Pinus radiata. We present production method, composition, and safety data from rat and dog toxicological and human clinical studies. The dry powder contains proanthocyanidins
Mijun Peng et al.
Environmental toxicology and pharmacology, 33(2), 327-333 (2012-02-04)
The effect of heavy metal ions, Cd(2+), Hg(2+) and Pb(2+) on taxifolin binding to bovine serum albumin (BSA) has been investigated by spectroscopic methods. The results indicated that the presence of heavy metal ions significantly affected the binding modes and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service