Skip to Content
Merck

Skip To

02430

Ethanesulfonic acid sodium salt monohydrate

≥98.0% (T)

Synonym(s):

Sodium-ethanesulfonate monohydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
10 g
Check Cart for Availability
€ 166,00

About This Item

Linear Formula:
CH3CH2SO3Na·H2O
CAS Number:
Molecular Weight:
150.13
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
12352100
EC Number:
226-194-9
MDL number:
Assay:
≥98.0% (T)

€ 166,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

anionic

Quality Level

assay

≥98.0% (T)

mp

248-256 °C (lit.)

functional group

sulfonic acid

SMILES string

[Na+].[H]O[H].CCS([O-])(=O)=O

InChI

1S/C2H6O3S.Na.H2O/c1-2-6(3,4)5;;/h2H2,1H3,(H,3,4,5);;1H2/q;+1;/p-1

InChI key

ZWDIDBOOWLDXON-UHFFFAOYSA-M


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.063521.0634672565
assay

≥98.0% (T)

assay

≥99.0% (alkalimetric)

assay

99.0-102.0% (alkalimetric)

assay

≥98.0% (KT)

Quality Level

200

Quality Level

300

Quality Level

300

Quality Level

200

mp

248-256 °C (lit.)

mp

183 °C (decomposition)

mp

-

mp

>300 °C (lit.)

description

anionic

description

-

description

-

description

-

functional group

sulfonic acid

functional group

-

functional group

-

functional group

amine


Still not finding the right product?


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Krzysztof M Bernadzki et al.
Frontiers in molecular neuroscience, 13, 104-104 (2020-06-27)
The neuromuscular junctions (NMJs) connect muscle fibers with motor neurons and enable the coordinated contraction of skeletal muscles. The dystrophin-associated glycoprotein complex (DGC) is an essential component of the postsynaptic machinery of the NMJ and is important for the maintenance
Haichao Wei et al.
Cell reports, 34(5), 108721-108721 (2021-02-04)
Spinal cord injury (SCI) is one of the most devastating neural injuries without effective therapeutic solutions. Astrocytes are the predominant component of the scar. Understanding the complex contributions of reactive astrocytes to SCI pathophysiologies is fundamentally important for developing therapeutic



Global Trade Item Number

SKUGTIN
Y000146004061837584596
PHR1003-1G04061835236794
1044006-500MG04061838671769
A5376-100G04061835542253
A5376-500G04061835542260
A5376-1KG04061833375808
A5376-250G04061833375815
A2093-100G04061833344583
A2093-500G04061833344606
A020000004061833327104
BP61704061834337454
02430-10G04061838617231

Questions

Reviews

No rating value

Active Filters