Skip to Content
Merck
All Photos(1)

Documents

18-1605

Sigma-Aldrich

Lithium perchlorate

SAJ special grade, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiClO4
CAS Number:
Molecular Weight:
106.39
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

SAJ special grade

Assay

≥99.0%

form

crystalline

reaction suitability

reagent type: oxidant

availability

available only in Japan

pH

6.0-9.5 (25 °C, 106.4 g/L)

mp

236 °C (lit.)

SMILES string

[Li+].[O-]Cl(=O)(=O)=O

InChI

1S/ClHO4.Li/c2-1(3,4)5;/h(H,2,3,4,5);/q;+1/p-1

InChI key

MHCFAGZWMAWTNR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Ox. Sol. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francisco Palacios et al.
The Journal of organic chemistry, 67(7), 2131-2135 (2002-04-02)
Functionalized keto-enamines 6 were obtained by nucleophilic addition of enol ethers to the imine moiety of 2-azadienes derived from dehydroaspartic esters 4. Reactions of 2-azadiene 4c containing three electron-withdrawing substituents (CO(2)R) with enol ethers 5 in the presence of lithium
Witold Darlewski et al.
Journal of hazardous materials, 175(1-3), 460-467 (2009-11-17)
A process of dibutyl sulphide (DBS) electro-oxidation using electrolysis and cyclic voltamperometry was investigated in water-methanol solution using different electrodes (platinum, boron doped diamond, graphite and glassy carbon). Obtained results indicate that the DBS electro-oxidation process is irreversible in voltamperometric
Shokaku Kim et al.
Organic letters, 4(21), 3735-3737 (2002-10-12)
[reaction: see text] N-Acyliminium cation of prolines was efficiently generated to accumulate in an undivided cell at 0 degrees C by an anodic oxidation of N-acylprolines or alpha'-phenylsulfanylated N-acylproline derivatives in a lithium perchlorate/nitromethane solution. The iminium cation intermediates gave
S Selvasekarapandian et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 65(5), 1234-1240 (2006-07-11)
The polymer electrolytes composed of poly(vinyl acetate) (PVAc) with various stoichiometric ratios of lithium perchlorate (LiClO(4)) salt have been prepared by solution casting method. The techniques Fourier transform infra-red (FTIR) and Laser Raman spectroscopy have been used to monitor polymer-salt
Increased T(c) in electrolyte-gated cuprates.
Anoop Singh Dhoot et al.
Advanced materials (Deerfield Beach, Fla.), 22(23), 2529-2533 (2010-05-07)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service