Skip to Content
Merck

Skip To

TMTP14250

Polycarbonate Membrane Filter, 5.0 μm Pore Size

Isopore, filter diam. 142 mm, hydrophilic

Synonym(s):

Isopore Membrane Filter

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
50 filters

Available to ship TODAYfromKuehne + Nagel Sp. z o.o.

2.330,00 PLN

About This Item

UNSPSC Code:
40161507
NACRES:
NB.24
eCl@ss:
32031602
Pore size:
5-20 % porosity, 5.0 μm pore size
Material:
plain filter, polycarbonate membrane, white filter
Feature:
hydrophilic
Diam.:
142 mm
Sterility:
non-sterile

2.330,00 PLN


Available to ship TODAYDetails


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

plain filter, polycarbonate membrane, white filter

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

Isopore, Millipore

parameter

140 °C max. temp., 250 mL/min-cm2-psi water flow rate (maximum flow rate per 1.0 psi), 50 L/min-cm2-psi air flow rate, ~550 mL/min-cm2 water flow rate at 10 psi (typical results)

diam.

142 mm

filter diam.

142 mm

thickness

20 μm

gravimetric extractables

<1%

refractive index

n/D 1.6

matrix

Isopore

pore size

5-20 % porosity, 5.0 μm pore size

shipped in

ambient

General description

Filter Code: TMTP
Filter Type: Screen filter
The Isopore membrane is a polycarbonate, track-etched screen filter recommended for all analyses in which the sample is viewed on the surface of the membrane. Isopore membrane offers distinct advantages for the analysis of airborne contaminants and other particles using optical or electron microscopy. The Isopore membrane is composed of polycarbonate film, which has a smooth, glass-like surface for clearer sample observation. The unique manufacturing process of the membrane ensures a precise pore diameter and a consistent pore size for accurate separation of samples by size. Matched-weight filters are not usually required because of low, constant tar and ash weights.

Features & Benefits:
•Membrane structure retains particles on the surface, simplifying counting and analysis
•Isopore membranes do not stain, resulting in low background interference
•Non-hygroscopic, allowing for rapid drying and reduced sample analysis time
•Translucent material does not require clearing for transmitted light microscopy; also available in brown variety

Applications:
Air Monitoring, Epifluorescent Microscopy, Chemotaxis Assays

Application

  • Parasitology
  • Chemotaxis
  • Bioassays
  • Cytology
  • Air monitoring
  • Microplastics analysis

Legal Information

ISOPORE is a trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Possible reduction in flow rates may occur after autoclaving.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
TMTP04700TMTP09030TMTP02500
material

plain filter, white filter, polycarbonate membrane

material

plain filter, polycarbonate membrane, white filter

material

plain filter, polycarbonate membrane, white filter

material

plain filter, polycarbonate membrane, white filter

parameter

140 °C max. temp., 50 L/min-cm2-psi air flow rate, 250 mL/min-cm2-psi water flow rate (maximum flow rate per 1.0 psi), ~550 mL/min-cm2 water flow rate at 10 psi (typical results)

parameter

140 °C max. temp., 250 mL/min-cm2-psi water flow rate (maximum flow rate per 1.0 psi), 50 L/min-cm2-psi air flow rate, ~550 mL/min-cm2 water flow rate at 10 psi (typical results)

parameter

140 °C max. temp., 250 mL/min-cm2-psi water flow rate (maximum flow rate per 1.0 psi), 50 L/min-cm2-psi air flow rate, ~550 mL/min-cm2 water flow rate at 10 psi (typical results)

parameter

140 °C max. temp., 250 mL/min-cm2-psi water flow rate (maximum flow rate per 1.0 psi), 50 L/min-cm2-psi air flow rate, 550 mL/min-cm2 water flow rate (typical flow rate at 10 psi)

diam.

142 mm

diam.

47 mm

diam.

90 mm

diam.

25 mm

manufacturer/tradename

Isopore, Millipore

manufacturer/tradename

Isopore, Millipore

manufacturer/tradename

Isopore, Millipore

manufacturer/tradename

Isopore, Millipore

pore size

5-20 % porosity, 5.0 μm pore size

pore size

5-20 % porosity, 5.0 μm pore size

pore size

5-20 % porosity, 5.0 μm pore size

pore size

5-20 % porosity, 5.0 μm pore size

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile


Still not finding the right product?




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters