Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 100 μg | Check Cart for Availability | € 453,00 |
About This Item
€ 453,00
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
5B8.2, monoclonal
species reactivity
human
concentration
1 mg/mL
technique(s)
immunohistochemistry: suitable, western blot: suitable
isotype
IgMκ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... SCARB1(949)
General description
Immunogen
Application
Apoptosis & Cancer
Apoptosis - Additional
Physical form
Preparation Note
Analysis Note
Western Blotting Analysis: 1.0 µg/mL of this antibody detected SCARB1 in 10 µg of human adrenal tissue lysate.
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| antibody form purified immunoglobulin | antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| species reactivity human | species reactivity rat, human, mouse | species reactivity human | species reactivity human |
| clone 5B8.2, monoclonal | clone polyclonal | clone polyclonal | clone polyclonal |
| biological source mouse | biological source rabbit | biological source rabbit | biological source rabbit |
| Gene Information human ... SCARB1(949) | Gene Information human ... SCARB1(949) | Gene Information human ... SCARB1(949) | Gene Information human ... SCARB1(949) |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| MABC730 | 04053252975608 |



