Skip to Content
Merck

Skip To

MABC161

Anti-BRCA1 Antibody, clone MS13

clone MS13, from mouse

Synonym(s):

Breast cancer type 1 susceptibility protein, RING finger protein 53

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
MS13, monoclonal
Application:
ICC, IHC, WB
Citations:
2
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MS13, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable, immunohistochemistry: suitable, western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BRCA1(672)

General description

Breast cancer type 1 susceptibility protein (BCRA1; RING finger protein 53) functions as a tumor suppressor. BRCA1 is an important member of the DNA repair pathway. After DNA damage, BRCA1 is phosphorylated on multiple serine residues by the ATR kinase or Chk2, and is localized to lesion sites with PCNA. BRCA1 interacts with a wide range of proteins including Rad50, NBS1, and Mre11, c-Abl tyrosine kinase, and γH2A.X, involved in the detection of damaged DNA and activation of appropriate repair pathways. BRCA1 may also respond to DNA damage by promoting gene expression through association with transcriptional proteins and RNA polymerase II, and may regulate the p53 pathway by this mechanism. Previous studies have indicated that mutations in the BRCA1 gene may contribute to the development of breast and ovarian cancer.
~220 kDa observed. The calculated molecular weight of this protein is 208 kDa, but can be observed at ~220 kDa due to high glycosylation. An uncharacterized band appears at ~65 kDa in some lysates.

Immunogen

Recombinant protein corresponding to human BRCA1.

Application

Anti-BRCA1 Antibody, clone MS13 detects level of BRCA1 & has been published & validated for use in Western Blotting, IHC, ICC.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected BRCA1 in human breast cancer and human ovarian cancer tissues.

Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected BRCA1 in HeLa and A431 cells.

Physical form

Format: Purified

Analysis Note

Evaluated by Western Blot in HeLa nuclear extract.

Western Blot Analysis: 0.5 µg/mL of this antibody detected BRCA1 on 10 µg of HeLa nuclear extract.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MABC199SAB4300490SAB5700340
Gene Information

human ... BRCA1(672)

Gene Information

human ... BRCA1(672)

Gene Information

human ... BRCA1(672)

Gene Information

human ... BRCA1(672)

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

-

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

rat, human, mouse

clone

MS13, monoclonal

clone

MS110, monoclonal

clone

polyclonal

clone

polyclonal

biological source

mouse

biological source

mouse

biological source

rabbit

biological source

rabbit


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters