Skip to Content
Merck
All Photos(1)

Documents

GF050

Sigma-Aldrich

Leptin, mouse

Leptin (Anti-Obesity Protein) is the protein product of the ob (obese) gene in mice & appears to be involved in appetite control.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.77

biological source

mouse

Quality Level

Assay

>95% (SDS-PAGE and HPLC)

manufacturer/tradename

Chemicon®

technique(s)

cell culture | mammalian: suitable

input

sample type mesenchymal stem cell(s)

UniProt accession no.

shipped in

wet ice

General description

Leptin (Anti-Obesity Protein) is the protein product of the ob (obese) gene in mice and appears to be involved in appetite control. The ob gene is expressed in adipose tissue and is thought to regulate the body′s fat stores. Mice with the ob/ob genotype also develop a form of diabetes similar to type II (non-insulin dependent) diabetes. Mouse leptin is a 16 kDa protein containing 147 amino acid residues.
Product Source: E.coli

Physical form

Lyophilized from 0.1% TFA, no preservatives.

Storage and Stability

The lyophilized powder, though stable at room temperature, is best stored desiccated below 0°C for up to 12 months from date of receipt. After a quick spin, reconstitute the mouse leptin in sterile distilled water to a concentration of 1.0-5.0 mg/mL. After reconstitution maintain at -20°C in undiluted aliquots for up to six months.

Analysis Note

Specific Activity: GF050 has been shown to be biologically active in two different mouse obesity models, ob/ob and NZO. Both strains of mice were treated via intraperitoneal injection once daily at a dose of 5 μg leptin/gm of body weight for 14 days. Significant effects on body weight, food consumption, and plasma glucose levels were observed compared to saline-treated controls.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service